Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46298685-46299177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
UniGene
Hs.420830
Gene Chromosome Location
Chr.19: 46297046-46343433 on Build GRCh38
Transcripts
NM_022462.4 NM_152795.3 NM_152796.2 XM_005259152.4 XM_005259155.4 XM_005259156.4 XM_017027132.1 XM_017027133.1 XM_017027134.1 XM_017027136.1 XM_017027138.1 XM_017027139.1 XM_017027140.1 XM_017027142.1

SNP Detail

SNP ID Chromosome Location
rs760033638 Chr.19: 46298923-46298923 on Build GRCh38
rs149233194 Chr.19: 46299043-46299043 on Build GRCh38
rs774030835 Chr.19: 46298791-46298791 on Build GRCh38
rs758896936 Chr.19: 46299076-46299076 on Build GRCh38
rs138386020 Chr.19: 46298904-46298904 on Build GRCh38
rs187327314 Chr.19: 46299130-46299130 on Build GRCh38
rs964998631 Chr.19: 46299154-46299154 on Build GRCh38
rs576129719 Chr.19: 46298880-46298880 on Build GRCh38
rs573709083 Chr.19: 46298838-46298838 on Build GRCh38
rs567968775 Chr.19: 46298870-46298870 on Build GRCh38
rs375834909 Chr.19: 46299110-46299110 on Build GRCh38
rs572140112 Chr.19: 46298896-46298896 on Build GRCh38
rs1043320459 Chr.19: 46298730-46298730 on Build GRCh38
rs979213300 Chr.19: 46299147-46299147 on Build GRCh38
rs954699553 Chr.19: 46299104-46299104 on Build GRCh38
rs1056703005 Chr.19: 46298955-46298955 on Build GRCh38
rs960362388 Chr.19: 46299109-46299109 on Build GRCh38
rs1051148906 Chr.19: 46298952-46298952 on Build GRCh38
rs549085185 Chr.19: 46299090-46299090 on Build GRCh38
rs966482270 Chr.19: 46298886-46298886 on Build GRCh38
rs887309937 Chr.19: 46298961-46298961 on Build GRCh38
rs979352044 Chr.19: 46298887-46298887 on Build GRCh38
rs1001152293 Chr.19: 46298787-46298787 on Build GRCh38
rs144508125 Chr.19: 46299068-46299068 on Build GRCh38
rs905495646 Chr.19: 46298744-46298744 on Build GRCh38
rs1004384502 Chr.19: 46298963-46298963 on Build GRCh38
rs545279821 Chr.19: 46298890-46298890 on Build GRCh38
rs35402294 Chr.19: 46298761-46298762 on Build GRCh38
rs993897412 Chr.19: 46298844-46298844 on Build GRCh38
rs887236370 Chr.19: 46298734-46298734 on Build GRCh38
rs111586797 Chr.19: 46299118-46299118 on Build GRCh38
rs1043655687 Chr.19: 46298899-46298899 on Build GRCh38
rs1004479325 Chr.19: 46298750-46298750 on Build GRCh38
rs947581624 Chr.19: 46298897-46298897 on Build GRCh38
rs994289220 Chr.19: 46298991-46298991 on Build GRCh38
rs534286759 Chr.19: 46298895-46298895 on Build GRCh38
rs998118787 Chr.19: 46299099-46299099 on Build GRCh38
rs764534084 Chr.19: 46299089-46299089 on Build GRCh38
rs28564298 Chr.19: 46298865-46298865 on Build GRCh38
rs1017192524 Chr.19: 46298757-46298757 on Build GRCh38
rs974730291 Chr.19: 46298892-46298892 on Build GRCh38
rs796675505 Chr.19: 46298897-46298897 on Build GRCh38
rs925229004 Chr.19: 46299150-46299150 on Build GRCh38
rs898730371 Chr.19: 46299047-46299047 on Build GRCh38
rs966554980 Chr.19: 46299129-46299129 on Build GRCh38
rs374145689 Chr.19: 46299101-46299101 on Build GRCh38
rs962409502 Chr.19: 46298786-46298786 on Build GRCh38
rs60593560 Chr.19: 46298889-46298889 on Build GRCh38
rs1005844099 Chr.19: 46298888-46298888 on Build GRCh38
rs1020652984 Chr.19: 46299119-46299119 on Build GRCh38
rs536240131 Chr.19: 46298852-46298852 on Build GRCh38
rs759696285 Chr.19: 46299108-46299108 on Build GRCh38

View Full Product Details