Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63104230-63104773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAR1A
Entrez Gene ID
768096
Gene Name
highly accelerated region 1A (non-protein coding)
Gene Aliases
HAR1F, LINC00064, NCRNA00064
UniGene
Hs.549606
Gene Chromosome Location
Chr.20: 63101292-63104385 on Build GRCh38
Transcripts
NR_003244.1

SNP Detail

SNP ID Chromosome Location
rs961171224 Chr.20: 63104619-63104619 on Build GRCh38
rs935020711 Chr.20: 63104752-63104752 on Build GRCh38
rs1019261969 Chr.20: 63104614-63104614 on Build GRCh38
rs140183532 Chr.20: 63104457-63104457 on Build GRCh38
rs557450565 Chr.20: 63104439-63104439 on Build GRCh38
rs368606267 Chr.20: 63104390-63104390 on Build GRCh38
rs1003049804 Chr.20: 63104522-63104522 on Build GRCh38
rs543941780 Chr.20: 63104656-63104656 on Build GRCh38
rs919730452 Chr.20: 63104425-63104425 on Build GRCh38
rs756711394 Chr.20: 63104721-63104721 on Build GRCh38
rs958329618 Chr.20: 63104271-63104271 on Build GRCh38
rs1011781972 Chr.20: 63104588-63104588 on Build GRCh38
rs568647604 Chr.20: 63104427-63104427 on Build GRCh38
rs996473917 Chr.20: 63104528-63104528 on Build GRCh38
rs973299446 Chr.20: 63104663-63104663 on Build GRCh38
rs990736617 Chr.20: 63104677-63104677 on Build GRCh38
rs562556808 Chr.20: 63104717-63104717 on Build GRCh38
rs1032891970 Chr.20: 63104260-63104260 on Build GRCh38
rs764894928 Chr.20: 63104387-63104387 on Build GRCh38
rs142505213 Chr.20: 63104503-63104503 on Build GRCh38
rs76803877 Chr.20: 63104474-63104474 on Build GRCh38
rs1046145384 Chr.20: 63104736-63104736 on Build GRCh38
rs533104030 Chr.20: 63104725-63104725 on Build GRCh38
rs879823285 Chr.20: 63104601-63104601 on Build GRCh38
rs891924935 Chr.20: 63104564-63104564 on Build GRCh38
rs947720603 Chr.20: 63104477-63104477 on Build GRCh38
rs1032531548 Chr.20: 63104501-63104501 on Build GRCh38
rs990601977 Chr.20: 63104299-63104300 on Build GRCh38
rs923167222 Chr.20: 63104414-63104414 on Build GRCh38
rs535378730 Chr.20: 63104312-63104312 on Build GRCh38
rs972876680 Chr.20: 63104652-63104652 on Build GRCh38
rs952647151 Chr.20: 63104657-63104657 on Build GRCh38
rs1026076454 Chr.20: 63104484-63104484 on Build GRCh38
rs902615368 Chr.20: 63104475-63104475 on Build GRCh38
rs539431882 Chr.20: 63104458-63104458 on Build GRCh38
rs561688679 Chr.20: 63104612-63104612 on Build GRCh38
rs999991857 Chr.20: 63104489-63104489 on Build GRCh38
rs906170012 Chr.20: 63104739-63104739 on Build GRCh38
rs752376052 Chr.20: 63104395-63104395 on Build GRCh38
rs935456650 Chr.20: 63104731-63104731 on Build GRCh38
rs777110883 Chr.20: 63104612-63104613 on Build GRCh38
rs916311764 Chr.20: 63104689-63104689 on Build GRCh38
rs540522216 Chr.20: 63104577-63104577 on Build GRCh38
rs551432671 Chr.20: 63104299-63104299 on Build GRCh38
rs933255652 Chr.20: 63104419-63104419 on Build GRCh38
rs895185681 Chr.20: 63104446-63104446 on Build GRCh38
rs765798050 Chr.20: 63104254-63104254 on Build GRCh38
rs916502760 Chr.20: 63104702-63104702 on Build GRCh38
rs1022125538 Chr.20: 63104609-63104609 on Build GRCh38
rs939730506 Chr.20: 63104393-63104393 on Build GRCh38
rs200892645 Chr.20: 63104613-63104613 on Build GRCh38
rs750699 Chr.20: 63104308-63104308 on Build GRCh38
rs758980811 Chr.20: 63104314-63104314 on Build GRCh38
rs1046396351 Chr.20: 63104724-63104724 on Build GRCh38
rs1054270104 Chr.20: 63104734-63104734 on Build GRCh38
rs986584697 Chr.20: 63104364-63104364 on Build GRCh38
rs535544355 Chr.20: 63104438-63104438 on Build GRCh38
rs1029244056 Chr.20: 63104568-63104568 on Build GRCh38
rs77772229 Chr.20: 63104653-63104653 on Build GRCh38
rs975381005 Chr.20: 63104274-63104274 on Build GRCh38

View Full Product Details