Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160003034-160003543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRN
Entrez Gene ID
100271873
Gene Name
antisense of IGF2R non-protein coding RNA
Gene Aliases
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088
UniGene
Hs.742166
Gene Chromosome Location
Chr.6: 160003291-160007664 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

SNP Detail

SNP ID Chromosome Location
rs186780532 Chr.6: 160003119-160003119 on Build GRCh38
rs112352185 Chr.6: 160003207-160003207 on Build GRCh38
rs969764381 Chr.6: 160003095-160003095 on Build GRCh38
rs896672151 Chr.6: 160003256-160003256 on Build GRCh38
rs957272702 Chr.6: 160003118-160003118 on Build GRCh38
rs966231099 Chr.6: 160003136-160003136 on Build GRCh38
rs887569790 Chr.6: 160003244-160003244 on Build GRCh38
rs1014142554 Chr.6: 160003360-160003360 on Build GRCh38
rs779872239 Chr.6: 160003436-160003436 on Build GRCh38
rs978713144 Chr.6: 160003381-160003381 on Build GRCh38
rs565907148 Chr.6: 160003313-160003313 on Build GRCh38
rs535344595 Chr.6: 160003377-160003377 on Build GRCh38
rs758242437 Chr.6: 160003301-160003301 on Build GRCh38
rs3798193 Chr.6: 160003082-160003082 on Build GRCh38
rs921980782 Chr.6: 160003169-160003169 on Build GRCh38
rs527342596 Chr.6: 160003238-160003238 on Build GRCh38
rs918200132 Chr.6: 160003073-160003073 on Build GRCh38
rs1024453785 Chr.6: 160003085-160003085 on Build GRCh38
rs1041423318 Chr.6: 160003197-160003197 on Build GRCh38
rs1032953045 Chr.6: 160003101-160003101 on Build GRCh38
rs750065751 Chr.6: 160003267-160003267 on Build GRCh38
rs897144469 Chr.6: 160003521-160003521 on Build GRCh38
rs529366953 Chr.6: 160003135-160003135 on Build GRCh38
rs956992109 Chr.6: 160003511-160003511 on Build GRCh38
rs756909509 Chr.6: 160003216-160003216 on Build GRCh38
rs993813735 Chr.6: 160003241-160003241 on Build GRCh38
rs1015040311 Chr.6: 160003060-160003060 on Build GRCh38
rs753333952 Chr.6: 160003183-160003183 on Build GRCh38
rs1010159718 Chr.6: 160003517-160003517 on Build GRCh38
rs746938615 Chr.6: 160003437-160003437 on Build GRCh38
rs1049031917 Chr.6: 160003187-160003187 on Build GRCh38
rs140079798 Chr.6: 160003246-160003246 on Build GRCh38
rs991868611 Chr.6: 160003363-160003363 on Build GRCh38
rs958991422 Chr.6: 160003061-160003061 on Build GRCh38
rs925463685 Chr.6: 160003384-160003384 on Build GRCh38
rs944110658 Chr.6: 160003479-160003479 on Build GRCh38
rs529939704 Chr.6: 160003446-160003446 on Build GRCh38
rs1014009698 Chr.6: 160003083-160003083 on Build GRCh38
rs976210958 Chr.6: 160003137-160003137 on Build GRCh38
rs1001188899 Chr.6: 160003096-160003096 on Build GRCh38
rs1032670229 Chr.6: 160003508-160003508 on Build GRCh38
rs959094281 Chr.6: 160003343-160003344 on Build GRCh38
rs769550589 Chr.6: 160003243-160003243 on Build GRCh38
rs1033406809 Chr.6: 160003312-160003312 on Build GRCh38
rs992839505 Chr.6: 160003062-160003062 on Build GRCh38

View Full Product Details