Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160004725-160005255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRN
Entrez Gene ID
100271873
Gene Name
antisense of IGF2R non-protein coding RNA
Gene Aliases
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088
UniGene
Hs.742166
Gene Chromosome Location
Chr.6: 160003291-160007664 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NR_047511.1 NR_047514.1 NM_000876.3

SNP Detail

SNP ID Chromosome Location
rs907214861 Chr.6: 160005196-160005196 on Build GRCh38
rs550896254 Chr.6: 160004873-160004873 on Build GRCh38
rs746116601 Chr.6: 160004976-160004976 on Build GRCh38
rs781247374 Chr.6: 160004849-160004849 on Build GRCh38
rs573877198 Chr.6: 160005119-160005119 on Build GRCh38
rs8191719 Chr.6: 160005176-160005176 on Build GRCh38
rs8191717 Chr.6: 160005157-160005157 on Build GRCh38
rs570436682 Chr.6: 160004872-160004872 on Build GRCh38
rs376514127 Chr.6: 160004886-160004886 on Build GRCh38
rs900799154 Chr.6: 160004859-160004859 on Build GRCh38
rs902762711 Chr.6: 160005061-160005061 on Build GRCh38
rs186547415 Chr.6: 160004878-160004878 on Build GRCh38
rs887293623 Chr.6: 160004875-160004875 on Build GRCh38
rs573917569 Chr.6: 160004748-160004748 on Build GRCh38
rs147580915 Chr.6: 160004808-160004808 on Build GRCh38
rs957988977 Chr.6: 160004806-160004806 on Build GRCh38
rs139221978 Chr.6: 160005195-160005195 on Build GRCh38
rs956604134 Chr.6: 160005013-160005013 on Build GRCh38
rs1017277690 Chr.6: 160005140-160005140 on Build GRCh38
rs8191715 Chr.6: 160004937-160004937 on Build GRCh38
rs747475054 Chr.6: 160005169-160005169 on Build GRCh38
rs145900439 Chr.6: 160005099-160005099 on Build GRCh38
rs915073511 Chr.6: 160005034-160005034 on Build GRCh38
rs550765918 Chr.6: 160004793-160004793 on Build GRCh38
rs564904856 Chr.6: 160004852-160004852 on Build GRCh38
rs1038584992 Chr.6: 160005152-160005152 on Build GRCh38
rs1047153981 Chr.6: 160005082-160005082 on Build GRCh38
rs908826537 Chr.6: 160005126-160005126 on Build GRCh38
rs937802072 Chr.6: 160005233-160005233 on Build GRCh38
rs976930967 Chr.6: 160004839-160004839 on Build GRCh38
rs534668411 Chr.6: 160004961-160004961 on Build GRCh38
rs375703653 Chr.6: 160004951-160004951 on Build GRCh38
rs989302601 Chr.6: 160005021-160005021 on Build GRCh38
rs8191716 Chr.6: 160004975-160004975 on Build GRCh38
rs565020420 Chr.6: 160005230-160005230 on Build GRCh38
rs545075379 Chr.6: 160004838-160004838 on Build GRCh38
rs938083288 Chr.6: 160004984-160004984 on Build GRCh38
rs942446400 Chr.6: 160004837-160004837 on Build GRCh38
rs922275345 Chr.6: 160005062-160005062 on Build GRCh38
rs1029841426 Chr.6: 160005081-160005081 on Build GRCh38
rs954316940 Chr.6: 160005090-160005090 on Build GRCh38
rs775724075 Chr.6: 160005165-160005165 on Build GRCh38
rs982402896 Chr.6: 160005211-160005211 on Build GRCh38
rs8191718 Chr.6: 160005171-160005171 on Build GRCh38
rs114504593 Chr.6: 160005035-160005035 on Build GRCh38
rs906599387 Chr.6: 160004758-160004758 on Build GRCh38
rs998797271 Chr.6: 160005071-160005071 on Build GRCh38
rs565649650 Chr.6: 160004956-160004956 on Build GRCh38
rs1037594135 Chr.6: 160004881-160004881 on Build GRCh38
rs1038526758 Chr.6: 160004797-160004797 on Build GRCh38
rs142083341 Chr.6: 160004874-160004874 on Build GRCh38
rs982009888 Chr.6: 160004772-160004772 on Build GRCh38
rs1013424483 Chr.6: 160005170-160005170 on Build GRCh38
rs985810308 Chr.6: 160004868-160004868 on Build GRCh38
rs552544097 Chr.6: 160005000-160005000 on Build GRCh38
rs768892523 Chr.6: 160005234-160005234 on Build GRCh38
rs893826598 Chr.6: 160005019-160005019 on Build GRCh38
rs1021651032 Chr.6: 160005045-160005045 on Build GRCh38
rs1021190202 Chr.6: 160004810-160004810 on Build GRCh38
rs8191714 Chr.6: 160004801-160004801 on Build GRCh38
rs769750274 Chr.6: 160005078-160005078 on Build GRCh38

View Full Product Details