Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 54202025-54202546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPVC1
Entrez Gene ID
3262
Gene Name
human papillomavirus (type 18) E5 central sequence-like 1
Gene Aliases
HPV18E5L, PE5L
UniGene
Hs.742598
Gene Chromosome Location
Chr.7: 54201224-54202421 on Build GRCh38
Transcripts
NR_004422.2

SNP Detail

SNP ID Chromosome Location
rs899824743 Chr.7: 54202105-54202105 on Build GRCh38
rs753380235 Chr.7: 54202154-54202154 on Build GRCh38
rs1047265864 Chr.7: 54202430-54202430 on Build GRCh38
rs191205877 Chr.7: 54202502-54202502 on Build GRCh38
rs969508093 Chr.7: 54202240-54202240 on Build GRCh38
rs1035169955 Chr.7: 54202510-54202510 on Build GRCh38
rs55746910 Chr.7: 54202497-54202497 on Build GRCh38
rs983021939 Chr.7: 54202389-54202389 on Build GRCh38
rs572292671 Chr.7: 54202062-54202062 on Build GRCh38
rs73132209 Chr.7: 54202106-54202106 on Build GRCh38
rs1040871181 Chr.7: 54202340-54202340 on Build GRCh38
rs747321433 Chr.7: 54202479-54202479 on Build GRCh38
rs901025163 Chr.7: 54202372-54202372 on Build GRCh38
rs537987063 Chr.7: 54202455-54202455 on Build GRCh38
rs903788431 Chr.7: 54202114-54202114 on Build GRCh38
rs758051827 Chr.7: 54202367-54202367 on Build GRCh38
rs780575777 Chr.7: 54202429-54202429 on Build GRCh38
rs181975207 Chr.7: 54202235-54202235 on Build GRCh38
rs908715658 Chr.7: 54202439-54202439 on Build GRCh38
rs555645411 Chr.7: 54202414-54202418 on Build GRCh38
rs546282484 Chr.7: 54202065-54202065 on Build GRCh38
rs1050731350 Chr.7: 54202112-54202112 on Build GRCh38
rs1027619745 Chr.7: 54202357-54202357 on Build GRCh38
rs932621683 Chr.7: 54202473-54202473 on Build GRCh38
rs771614003 Chr.7: 54202144-54202144 on Build GRCh38
rs551588566 Chr.7: 54202277-54202277 on Build GRCh38
rs148079302 Chr.7: 54202259-54202259 on Build GRCh38
rs956793303 Chr.7: 54202258-54202258 on Build GRCh38
rs944246459 Chr.7: 54202448-54202448 on Build GRCh38
rs559490068 Chr.7: 54202245-54202245 on Build GRCh38
rs921362874 Chr.7: 54202470-54202470 on Build GRCh38
rs185159461 Chr.7: 54202495-54202495 on Build GRCh38
rs1024733183 Chr.7: 54202211-54202211 on Build GRCh38
rs756676244 Chr.7: 54202159-54202159 on Build GRCh38
rs535819511 Chr.7: 54202280-54202280 on Build GRCh38
rs570210576 Chr.7: 54202450-54202450 on Build GRCh38
rs868029477 Chr.7: 54202494-54202494 on Build GRCh38
rs562125346 Chr.7: 54202153-54202153 on Build GRCh38
rs1047230299 Chr.7: 54202052-54202052 on Build GRCh38
rs942654540 Chr.7: 54202329-54202329 on Build GRCh38
rs952949926 Chr.7: 54202381-54202381 on Build GRCh38
rs143604474 Chr.7: 54202182-54202182 on Build GRCh38
rs116029335 Chr.7: 54202092-54202092 on Build GRCh38
rs994352468 Chr.7: 54202134-54202134 on Build GRCh38
rs976771973 Chr.7: 54202457-54202457 on Build GRCh38
rs928149177 Chr.7: 54202241-54202241 on Build GRCh38
rs571622426 Chr.7: 54202129-54202129 on Build GRCh38
rs1041153935 Chr.7: 54202102-54202102 on Build GRCh38
rs982276133 Chr.7: 54202241-54202245 on Build GRCh38
rs771080652 Chr.7: 54202327-54202327 on Build GRCh38
rs1000672779 Chr.7: 54202116-54202116 on Build GRCh38
rs915214630 Chr.7: 54202272-54202272 on Build GRCh38
rs763980593 Chr.7: 54202509-54202509 on Build GRCh38
rs189368565 Chr.7: 54202110-54202110 on Build GRCh38

View Full Product Details