Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144110010-144110511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR97
Entrez Gene ID
340390
Gene Name
WD repeat domain 97
Gene Aliases
KIAA1875
UniGene
Hs.98723
Gene Chromosome Location
Chr.8: 144107692-144116392 on Build GRCh38
Transcripts
NM_001316309.1 XM_017013385.1 XM_017013386.1 XM_017013387.1 XM_017013388.1 XM_017013389.1 XM_017013390.1 XM_017013391.1 XM_017013392.1 XM_017013393.1 XM_017013394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603893 c.1726G>A p.D576N Chr.8: 144110139-144110139 on Build GRCh38
7261077 c.1784C>T p.A595V Chr.8: 144110197-144110197 on Build GRCh38
5601695 c.1841C>T p.S614L Chr.8: 144110254-144110254 on Build GRCh38
3747740 c.1910G>C p.R637P Chr.8: 144110407-144110407 on Build GRCh38
4963277 c.1968C>T p.T656T Chr.8: 144110465-144110465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907403514 Chr.8: 144110160-144110160 on Build GRCh38
rs939076066 Chr.8: 144110445-144110445 on Build GRCh38
rs930209150 Chr.8: 144110329-144110329 on Build GRCh38
rs529131932 Chr.8: 144110355-144110355 on Build GRCh38
rs1025602833 Chr.8: 144110309-144110309 on Build GRCh38
rs539883762 Chr.8: 144110384-144110384 on Build GRCh38
rs758813263 Chr.8: 144110116-144110116 on Build GRCh38
rs553396531 Chr.8: 144110168-144110168 on Build GRCh38
rs776325907 Chr.8: 144110289-144110289 on Build GRCh38
rs765603414 Chr.8: 144110287-144110287 on Build GRCh38
rs374296026 Chr.8: 144110217-144110217 on Build GRCh38
rs997356097 Chr.8: 144110408-144110408 on Build GRCh38
rs981692122 Chr.8: 144110338-144110338 on Build GRCh38
rs561801815 Chr.8: 144110397-144110397 on Build GRCh38
rs764575653 Chr.8: 144110383-144110383 on Build GRCh38
rs1044776938 Chr.8: 144110114-144110114 on Build GRCh38
rs564397000 Chr.8: 144110333-144110333 on Build GRCh38
rs759095614 Chr.8: 144110330-144110330 on Build GRCh38
rs928784687 Chr.8: 144110346-144110346 on Build GRCh38
rs985534786 Chr.8: 144110256-144110256 on Build GRCh38
rs779536021 Chr.8: 144110086-144110086 on Build GRCh38
rs965461444 Chr.8: 144110300-144110300 on Build GRCh38
rs755389178 Chr.8: 144110360-144110360 on Build GRCh38
rs911193782 Chr.8: 144110049-144110049 on Build GRCh38
rs866715148 Chr.8: 144110484-144110484 on Build GRCh38
rs772822285 Chr.8: 144110282-144110282 on Build GRCh38
rs541984381 Chr.8: 144110180-144110180 on Build GRCh38
rs781369729 Chr.8: 144110402-144110402 on Build GRCh38
rs868705777 Chr.8: 144110033-144110033 on Build GRCh38
rs764791935 Chr.8: 144110339-144110339 on Build GRCh38
rs953344530 Chr.8: 144110243-144110243 on Build GRCh38
rs910079759 Chr.8: 144110385-144110385 on Build GRCh38
rs918846689 Chr.8: 144110087-144110087 on Build GRCh38
rs998044343 Chr.8: 144110266-144110266 on Build GRCh38
rs967260772 Chr.8: 144110205-144110205 on Build GRCh38
rs114553997 Chr.8: 144110174-144110174 on Build GRCh38
rs969550336 Chr.8: 144110393-144110393 on Build GRCh38
rs946654583 Chr.8: 144110490-144110490 on Build GRCh38
rs918622902 Chr.8: 144110297-144110297 on Build GRCh38
rs556920881 Chr.8: 144110188-144110188 on Build GRCh38
rs1043614662 Chr.8: 144110225-144110225 on Build GRCh38
rs1030083903 Chr.8: 144110223-144110223 on Build GRCh38
rs145906584 Chr.8: 144110465-144110465 on Build GRCh38
rs1007003298 Chr.8: 144110291-144110291 on Build GRCh38
rs570465960 Chr.8: 144110095-144110095 on Build GRCh38
rs1019938383 Chr.8: 144110197-144110197 on Build GRCh38
rs1011179805 Chr.8: 144110194-144110194 on Build GRCh38
rs757059763 Chr.8: 144110054-144110079 on Build GRCh38
rs1007786079 Chr.8: 144110447-144110447 on Build GRCh38
rs902862123 Chr.8: 144110395-144110395 on Build GRCh38
rs143515279 Chr.8: 144110132-144110132 on Build GRCh38
rs781314541 Chr.8: 144110201-144110201 on Build GRCh38
rs888933565 Chr.8: 144110434-144110434 on Build GRCh38
rs768629709 Chr.8: 144110259-144110259 on Build GRCh38
rs546076298 Chr.8: 144110279-144110279 on Build GRCh38
rs866478837 Chr.8: 144110417-144110417 on Build GRCh38
rs551896269 Chr.8: 144110457-144110457 on Build GRCh38
rs745916344 Chr.8: 144110258-144110258 on Build GRCh38
rs758766664 Chr.8: 144110370-144110370 on Build GRCh38
rs1030678488 Chr.8: 144110425-144110425 on Build GRCh38
rs575598084 Chr.8: 144110264-144110264 on Build GRCh38
rs138869895 Chr.8: 144110467-144110467 on Build GRCh38
rs577629379 Chr.8: 144110485-144110485 on Build GRCh38
rs34523416 Chr.8: 144110196-144110197 on Build GRCh38
rs998600809 Chr.8: 144110306-144110306 on Build GRCh38
rs761611511 Chr.8: 144110267-144110267 on Build GRCh38
rs955928180 Chr.8: 144110084-144110084 on Build GRCh38
rs1001252462 Chr.8: 144110171-144110171 on Build GRCh38
rs965353262 Chr.8: 144110071-144110071 on Build GRCh38
rs976755733 Chr.8: 144110080-144110080 on Build GRCh38
rs746037909 Chr.8: 144110460-144110460 on Build GRCh38
rs927727602 Chr.8: 144110410-144110410 on Build GRCh38
rs941373938 Chr.8: 144110053-144110053 on Build GRCh38
rs371576441 Chr.8: 144110416-144110416 on Build GRCh38
rs930238642 Chr.8: 144110097-144110097 on Build GRCh38
rs1041362963 Chr.8: 144110386-144110386 on Build GRCh38
rs760435691 Chr.8: 144110286-144110286 on Build GRCh38
rs984696507 Chr.8: 144110094-144110094 on Build GRCh38
rs951626830 Chr.8: 144110089-144110089 on Build GRCh38
rs1038607176 Chr.8: 144110077-144110077 on Build GRCh38

View Full Product Details