Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 114639337-114639841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR2C
Entrez Gene ID
3358
Gene Name
5-hydroxytryptamine receptor 2C
Gene Aliases
5-HT1C, 5-HT2C, 5-HTR2C, 5HTR2C, HTR1C
UniGene
Hs.149037
Gene Chromosome Location
Chr.X: 114584078-114910061 on Build GRCh38
Transcripts
NM_000868.3 NM_001256760.2 NM_001256761.2 NR_031581.1

Gene Symbol
MIR764
Entrez Gene ID
100313838
Gene Name
microRNA 764
Gene Aliases
hsa-mir-764
UniGene
Gene Chromosome Location
Chr.X: 114639435-114639519 on Build GRCh38
Transcripts
NM_000868.3 NM_001256760.2 NM_001256761.2 NR_031581.1

SNP Detail

SNP ID Chromosome Location
rs782792145 Chr.X: 114639792-114639792 on Build GRCh38
rs903763439 Chr.X: 114639780-114639780 on Build GRCh38
rs112857617 Chr.X: 114639665-114639665 on Build GRCh38
rs942375648 Chr.X: 114639763-114639763 on Build GRCh38
rs782217809 Chr.X: 114639565-114639565 on Build GRCh38
rs76971695 Chr.X: 114639531-114639531 on Build GRCh38
rs782334116 Chr.X: 114639546-114639546 on Build GRCh38
rs782371045 Chr.X: 114639396-114639396 on Build GRCh38
rs368383537 Chr.X: 114639525-114639525 on Build GRCh38
rs782593861 Chr.X: 114639367-114639367 on Build GRCh38
rs78602960 Chr.X: 114639571-114639571 on Build GRCh38
rs782603380 Chr.X: 114639544-114639544 on Build GRCh38
rs782779390 Chr.X: 114639415-114639415 on Build GRCh38
rs1039636528 Chr.X: 114639778-114639778 on Build GRCh38
rs1050784052 Chr.X: 114639733-114639733 on Build GRCh38
rs782040023 Chr.X: 114639526-114639526 on Build GRCh38
rs781904343 Chr.X: 114639450-114639450 on Build GRCh38
rs929690150 Chr.X: 114639713-114639713 on Build GRCh38
rs781977122 Chr.X: 114639628-114639628 on Build GRCh38
rs782750814 Chr.X: 114639572-114639572 on Build GRCh38
rs192237597 Chr.X: 114639547-114639547 on Build GRCh38
rs782258540 Chr.X: 114639391-114639391 on Build GRCh38
rs377648254 Chr.X: 114639464-114639464 on Build GRCh38
rs782722555 Chr.X: 114639529-114639529 on Build GRCh38
rs77486158 Chr.X: 114639362-114639362 on Build GRCh38
rs782658385 Chr.X: 114639743-114639743 on Build GRCh38
rs918351139 Chr.X: 114639674-114639674 on Build GRCh38

View Full Product Details