Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 12406885-12407427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG2P1
Entrez Gene ID
352887
Gene Name
glycogenin 2 pseudogene 1
Gene Aliases
GYG2P
UniGene
Hs.546221
Gene Chromosome Location
Chr.Y: 12406116-12421590 on Build GRCh38
Transcripts
NR_033667.1

SNP Detail

SNP ID Chromosome Location
rs772238636 Chr.Y: 12407333-12407333 on Build GRCh38
rs758654262 Chr.Y: 12407046-12407046 on Build GRCh38
rs756848289 Chr.Y: 12407226-12407226 on Build GRCh38
rs746698292 Chr.Y: 12406972-12406972 on Build GRCh38
rs764798557 Chr.Y: 12407324-12407324 on Build GRCh38

View Full Product Details