Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43262016-43262563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPKP3
Entrez Gene ID
399881
Gene Name
heterogeneous nuclear ribonucleoprotein K pseudogene 3
Gene Aliases
-
UniGene
Hs.446761
Gene Chromosome Location
Chr.11: 43261504-43269369 on Build GRCh38
Transcripts
NR_033868.1

SNP Detail

SNP ID Chromosome Location
rs776121727 Chr.11: 43262055-43262056 on Build GRCh38
rs201499316 Chr.11: 43262056-43262057 on Build GRCh38
rs1022772240 Chr.11: 43262294-43262294 on Build GRCh38
rs1002497273 Chr.11: 43262400-43262400 on Build GRCh38
rs911480505 Chr.11: 43262442-43262442 on Build GRCh38
rs377012965 Chr.11: 43262067-43262068 on Build GRCh38
rs12361593 Chr.11: 43262427-43262427 on Build GRCh38
rs577539819 Chr.11: 43262232-43262232 on Build GRCh38
rs536059296 Chr.11: 43262116-43262116 on Build GRCh38
rs143255343 Chr.11: 43262359-43262359 on Build GRCh38
rs1015733849 Chr.11: 43262151-43262151 on Build GRCh38
rs185400907 Chr.11: 43262408-43262408 on Build GRCh38
rs961538279 Chr.11: 43262171-43262172 on Build GRCh38
rs569918230 Chr.11: 43262043-43262043 on Build GRCh38
rs954441135 Chr.11: 43262202-43262202 on Build GRCh38
rs879074565 Chr.11: 43262056-43262056 on Build GRCh38
rs985684672 Chr.11: 43262529-43262529 on Build GRCh38
rs180902406 Chr.11: 43262347-43262347 on Build GRCh38
rs867009713 Chr.11: 43262252-43262252 on Build GRCh38
rs866981159 Chr.11: 43262343-43262343 on Build GRCh38
rs939021000 Chr.11: 43262535-43262535 on Build GRCh38
rs942829064 Chr.11: 43262449-43262449 on Build GRCh38
rs573674203 Chr.11: 43262088-43262088 on Build GRCh38
rs1043947146 Chr.11: 43262363-43262363 on Build GRCh38
rs752539893 Chr.11: 43262164-43262164 on Build GRCh38
rs1015662060 Chr.11: 43262478-43262478 on Build GRCh38
rs763249666 Chr.11: 43262115-43262115 on Build GRCh38
rs528615681 Chr.11: 43262499-43262499 on Build GRCh38
rs894940464 Chr.11: 43262187-43262187 on Build GRCh38
rs1055052231 Chr.11: 43262405-43262405 on Build GRCh38
rs149967829 Chr.11: 43262485-43262485 on Build GRCh38
rs542841263 Chr.11: 43262284-43262284 on Build GRCh38
rs1040115856 Chr.11: 43262329-43262329 on Build GRCh38
rs139251976 Chr.11: 43262124-43262124 on Build GRCh38
rs1034811638 Chr.11: 43262172-43262172 on Build GRCh38
rs974282614 Chr.11: 43262483-43262483 on Build GRCh38
rs537206837 Chr.11: 43262048-43262048 on Build GRCh38
rs892278856 Chr.11: 43262053-43262053 on Build GRCh38
rs16937425 Chr.11: 43262525-43262525 on Build GRCh38
rs930437898 Chr.11: 43262513-43262513 on Build GRCh38
rs867694290 Chr.11: 43262317-43262317 on Build GRCh38
rs1013585702 Chr.11: 43262130-43262130 on Build GRCh38
rs1056274145 Chr.11: 43262538-43262538 on Build GRCh38
rs968180267 Chr.11: 43262297-43262297 on Build GRCh38
rs955602886 Chr.11: 43262373-43262373 on Build GRCh38
rs207471855 Chr.11: 43262412-43262412 on Build GRCh38
rs545327387 Chr.11: 43262250-43262250 on Build GRCh38
rs866389783 Chr.11: 43262508-43262508 on Build GRCh38
rs16937423 Chr.11: 43262498-43262498 on Build GRCh38
rs920215907 Chr.11: 43262181-43262181 on Build GRCh38
rs552916212 Chr.11: 43262184-43262184 on Build GRCh38
rs896468204 Chr.11: 43262411-43262411 on Build GRCh38
rs1043677196 Chr.11: 43262076-43262076 on Build GRCh38
rs575553167 Chr.11: 43262276-43262276 on Build GRCh38
rs555209059 Chr.11: 43262062-43262062 on Build GRCh38
rs563536472 Chr.11: 43262251-43262251 on Build GRCh38
rs886144481 Chr.11: 43262532-43262532 on Build GRCh38

View Full Product Details