Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43262832-43263382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPKP3
Entrez Gene ID
399881
Gene Name
heterogeneous nuclear ribonucleoprotein K pseudogene 3
Gene Aliases
-
UniGene
Hs.446761
Gene Chromosome Location
Chr.11: 43261504-43269369 on Build GRCh38
Transcripts
NR_033868.1

SNP Detail

SNP ID Chromosome Location
rs948027171 Chr.11: 43262861-43262861 on Build GRCh38
rs1032530797 Chr.11: 43263229-43263229 on Build GRCh38
rs77134638 Chr.11: 43262988-43262988 on Build GRCh38
rs1040106714 Chr.11: 43263341-43263341 on Build GRCh38
rs925200402 Chr.11: 43263266-43263266 on Build GRCh38
rs534867422 Chr.11: 43263186-43263186 on Build GRCh38
rs148593314 Chr.11: 43262967-43262967 on Build GRCh38
rs377536589 Chr.11: 43263119-43263119 on Build GRCh38
rs966490040 Chr.11: 43262905-43262905 on Build GRCh38
rs1043611685 Chr.11: 43263265-43263265 on Build GRCh38
rs765215318 Chr.11: 43263323-43263323 on Build GRCh38
rs1037003589 Chr.11: 43262985-43262985 on Build GRCh38
rs374338694 Chr.11: 43263292-43263292 on Build GRCh38
rs1019785945 Chr.11: 43263129-43263129 on Build GRCh38
rs866872806 Chr.11: 43263115-43263115 on Build GRCh38
rs1018315253 Chr.11: 43263013-43263013 on Build GRCh38
rs533195034 Chr.11: 43263073-43263073 on Build GRCh38
rs878998837 Chr.11: 43262943-43262943 on Build GRCh38
rs891202714 Chr.11: 43263322-43263322 on Build GRCh38
rs760465979 Chr.11: 43263222-43263222 on Build GRCh38
rs959114544 Chr.11: 43263256-43263256 on Build GRCh38
rs141832849 Chr.11: 43263176-43263176 on Build GRCh38
rs866678725 Chr.11: 43263248-43263248 on Build GRCh38
rs899792274 Chr.11: 43263356-43263356 on Build GRCh38
rs575588015 Chr.11: 43262932-43262932 on Build GRCh38
rs866473716 Chr.11: 43263144-43263144 on Build GRCh38
rs763195349 Chr.11: 43263150-43263150 on Build GRCh38
rs891230254 Chr.11: 43263001-43263001 on Build GRCh38
rs1007103161 Chr.11: 43263100-43263100 on Build GRCh38
rs911343279 Chr.11: 43263346-43263346 on Build GRCh38
rs10734513 Chr.11: 43262919-43262919 on Build GRCh38
rs771207503 Chr.11: 43263156-43263156 on Build GRCh38
rs975779592 Chr.11: 43262876-43262876 on Build GRCh38
rs753706152 Chr.11: 43263287-43263287 on Build GRCh38
rs559548281 Chr.11: 43263044-43263044 on Build GRCh38
rs748573575 Chr.11: 43263002-43263002 on Build GRCh38
rs375350149 Chr.11: 43262933-43262933 on Build GRCh38
rs889856895 Chr.11: 43263048-43263048 on Build GRCh38
rs544585340 Chr.11: 43263264-43263264 on Build GRCh38
rs531092385 Chr.11: 43263165-43263165 on Build GRCh38
rs951542309 Chr.11: 43263138-43263138 on Build GRCh38
rs1021004056 Chr.11: 43262891-43262891 on Build GRCh38
rs917716824 Chr.11: 43263289-43263289 on Build GRCh38
rs991774051 Chr.11: 43263225-43263225 on Build GRCh38
rs866639983 Chr.11: 43262930-43262930 on Build GRCh38
rs916284426 Chr.11: 43263263-43263263 on Build GRCh38
rs12276669 Chr.11: 43263132-43263132 on Build GRCh38
rs949298347 Chr.11: 43262983-43262983 on Build GRCh38
rs538667313 Chr.11: 43263293-43263293 on Build GRCh38
rs917875292 Chr.11: 43262965-43262965 on Build GRCh38
rs1020637819 Chr.11: 43263168-43263168 on Build GRCh38
rs964636873 Chr.11: 43263024-43263024 on Build GRCh38
rs561154050 Chr.11: 43262959-43262959 on Build GRCh38
rs1010452879 Chr.11: 43263163-43263163 on Build GRCh38
rs979568278 Chr.11: 43262917-43262917 on Build GRCh38
rs186008881 Chr.11: 43263065-43263065 on Build GRCh38
rs549376009 Chr.11: 43263167-43263167 on Build GRCh38
rs770477416 Chr.11: 43263021-43263021 on Build GRCh38
rs995812651 Chr.11: 43263116-43263116 on Build GRCh38
rs556981234 Chr.11: 43263343-43263343 on Build GRCh38
rs551356456 Chr.11: 43263087-43263087 on Build GRCh38
rs982923990 Chr.11: 43263169-43263169 on Build GRCh38

View Full Product Details