Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84509117-84509669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA6L5P
Entrez Gene ID
374650
Gene Name
golgin A6 family-like 5, pseudogene
Gene Aliases
GOLGA6L5, GOLGA6L8
UniGene
Hs.454625
Gene Chromosome Location
Chr.15: 84506168-84516847 on Build GRCh38
Transcripts
NR_003246.4

SNP Detail

SNP ID Chromosome Location
rs2436247 Chr.15: 84509358-84509358 on Build GRCh38
rs1006528139 Chr.15: 84509614-84509614 on Build GRCh38
rs1042815915 Chr.15: 84509537-84509537 on Build GRCh38
rs1053300161 Chr.15: 84509223-84509223 on Build GRCh38
rs889431602 Chr.15: 84509609-84509609 on Build GRCh38
rs62029636 Chr.15: 84509183-84509183 on Build GRCh38
rs1041328522 Chr.15: 84509627-84509627 on Build GRCh38
rs425743 Chr.15: 84509137-84509137 on Build GRCh38
rs913429052 Chr.15: 84509436-84509436 on Build GRCh38
rs879819185 Chr.15: 84509555-84509555 on Build GRCh38
rs571942 Chr.15: 84509481-84509481 on Build GRCh38
rs947129785 Chr.15: 84509521-84509521 on Build GRCh38
rs372022097 Chr.15: 84509473-84509473 on Build GRCh38
rs3877056 Chr.15: 84509359-84509359 on Build GRCh38
rs866888838 Chr.15: 84509588-84509588 on Build GRCh38
rs938553936 Chr.15: 84509200-84509200 on Build GRCh38

View Full Product Details