Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143559374-143559926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSBP5
Entrez Gene ID
441046
Gene Name
glucuronidase, beta pseudogene 5
Gene Aliases
GYPELOC441046
UniGene
Hs.135705
Gene Chromosome Location
Chr.4: 143559472-143561460 on Build GRCh38
Transcripts
NR_003675.2

SNP Detail

SNP ID Chromosome Location
rs1019566062 Chr.4: 143559777-143559777 on Build GRCh38
rs111306518 Chr.4: 143559623-143559623 on Build GRCh38
rs1008542303 Chr.4: 143559765-143559765 on Build GRCh38
rs571348991 Chr.4: 143559467-143559467 on Build GRCh38
rs372575548 Chr.4: 143559843-143559843 on Build GRCh38
rs940742705 Chr.4: 143559892-143559892 on Build GRCh38
rs917835007 Chr.4: 143559573-143559573 on Build GRCh38
rs564949750 Chr.4: 143559783-143559783 on Build GRCh38
rs1049333052 Chr.4: 143559494-143559494 on Build GRCh38
rs758286156 Chr.4: 143559673-143559673 on Build GRCh38
rs565327386 Chr.4: 143559880-143559880 on Build GRCh38
rs1019879345 Chr.4: 143559757-143559757 on Build GRCh38
rs1046007170 Chr.4: 143559683-143559683 on Build GRCh38
rs967328600 Chr.4: 143559421-143559421 on Build GRCh38
rs949211140 Chr.4: 143559472-143559472 on Build GRCh38
rs907886558 Chr.4: 143559679-143559679 on Build GRCh38
rs1044836000 Chr.4: 143559629-143559629 on Build GRCh38
rs1049049669 Chr.4: 143559674-143559674 on Build GRCh38
rs532637997 Chr.4: 143559460-143559460 on Build GRCh38
rs375420078 Chr.4: 143559864-143559864 on Build GRCh38
rs892848797 Chr.4: 143559739-143559739 on Build GRCh38
rs750668829 Chr.4: 143559425-143559425 on Build GRCh38
rs368644345 Chr.4: 143559795-143559795 on Build GRCh38
rs1039147418 Chr.4: 143559478-143559478 on Build GRCh38
rs184836379 Chr.4: 143559643-143559643 on Build GRCh38
rs912370716 Chr.4: 143559618-143559618 on Build GRCh38
rs964213314 Chr.4: 143559785-143559785 on Build GRCh38
rs1001675834 Chr.4: 143559521-143559521 on Build GRCh38
rs531385326 Chr.4: 143559714-143559714 on Build GRCh38
rs1010880918 Chr.4: 143559504-143559504 on Build GRCh38
rs534218599 Chr.4: 143559519-143559519 on Build GRCh38
rs929642329 Chr.4: 143559562-143559562 on Build GRCh38
rs191257689 Chr.4: 143559520-143559520 on Build GRCh38
rs114918376 Chr.4: 143559680-143559680 on Build GRCh38
rs896336445 Chr.4: 143559461-143559461 on Build GRCh38
rs886655406 Chr.4: 143559506-143559506 on Build GRCh38
rs532193507 Chr.4: 143559796-143559796 on Build GRCh38
rs553317598 Chr.4: 143559495-143559495 on Build GRCh38
rs1001746925 Chr.4: 143559720-143559720 on Build GRCh38
rs756351007 Chr.4: 143559458-143559458 on Build GRCh38
rs187696522 Chr.4: 143559863-143559863 on Build GRCh38
rs576914846 Chr.4: 143559792-143559792 on Build GRCh38
rs917827627 Chr.4: 143559845-143559845 on Build GRCh38
rs1026724811 Chr.4: 143559820-143559820 on Build GRCh38
rs780109767 Chr.4: 143559480-143559480 on Build GRCh38
rs769171544 Chr.4: 143559493-143559493 on Build GRCh38
rs574136307 Chr.4: 143559525-143559525 on Build GRCh38
rs922045044 Chr.4: 143559645-143559645 on Build GRCh38
rs17017911 Chr.4: 143559481-143559481 on Build GRCh38
rs186469787 Chr.4: 143559508-143559508 on Build GRCh38
rs369330726 Chr.4: 143559655-143559655 on Build GRCh38
rs928871452 Chr.4: 143559903-143559903 on Build GRCh38
rs544817342 Chr.4: 143559526-143559526 on Build GRCh38
rs1046912996 Chr.4: 143559459-143559459 on Build GRCh38
rs919563865 Chr.4: 143559561-143559561 on Build GRCh38
rs907363621 Chr.4: 143559709-143559709 on Build GRCh38
rs967100857 Chr.4: 143559760-143559760 on Build GRCh38
rs973748005 Chr.4: 143559552-143559552 on Build GRCh38
rs949269409 Chr.4: 143559862-143559863 on Build GRCh38
rs1033389747 Chr.4: 143559494-143559494 on Build GRCh38
rs868103376 Chr.4: 143559788-143559788 on Build GRCh38
rs930670362 Chr.4: 143559482-143559482 on Build GRCh38
rs371527234 Chr.4: 143559900-143559900 on Build GRCh38
rs748670498 Chr.4: 143559549-143559549 on Build GRCh38
rs142093305 Chr.4: 143559882-143559882 on Build GRCh38
rs561203648 Chr.4: 143559713-143559713 on Build GRCh38
rs549242813 Chr.4: 143559895-143559895 on Build GRCh38
rs986750036 Chr.4: 143559564-143559564 on Build GRCh38
rs146201727 Chr.4: 143559410-143559410 on Build GRCh38
rs578018617 Chr.4: 143559567-143559567 on Build GRCh38
rs1034103279 Chr.4: 143559522-143559522 on Build GRCh38
rs1018425158 Chr.4: 143559736-143559736 on Build GRCh38
rs984885484 Chr.4: 143559866-143559866 on Build GRCh38
rs967677562 Chr.4: 143559539-143559539 on Build GRCh38
rs891345211 Chr.4: 143559705-143559705 on Build GRCh38
rs1019910864 Chr.4: 143559527-143559527 on Build GRCh38
rs527735450 Chr.4: 143559646-143559646 on Build GRCh38
rs1055898609 Chr.4: 143559728-143559728 on Build GRCh38
rs556500374 Chr.4: 143559555-143559555 on Build GRCh38
rs963806629 Chr.4: 143559762-143559764 on Build GRCh38
rs987000083 Chr.4: 143559423-143559423 on Build GRCh38
rs944760106 Chr.4: 143559437-143559437 on Build GRCh38
rs1046293356 Chr.4: 143559518-143559518 on Build GRCh38
rs549922081 Chr.4: 143559751-143559751 on Build GRCh38
rs951156288 Chr.4: 143559828-143559828 on Build GRCh38
rs1054478966 Chr.4: 143559647-143559647 on Build GRCh38

View Full Product Details