Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 189473404-189473966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90AA4P
Entrez Gene ID
3323
Gene Name
heat shock protein 90 alpha family class A member 4, pseudogene
Gene Aliases
HSP90Ad, HSPCAL2
UniGene
Hs.553481
Gene Chromosome Location
Chr.4: 189472906-189475853 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs546739730 Chr.4: 189473659-189473659 on Build GRCh38
rs772167590 Chr.4: 189473453-189473453 on Build GRCh38
rs752528240 Chr.4: 189473876-189473876 on Build GRCh38
rs144103136 Chr.4: 189473790-189473790 on Build GRCh38
rs917001875 Chr.4: 189473557-189473557 on Build GRCh38
rs755937181 Chr.4: 189473909-189473909 on Build GRCh38
rs767407995 Chr.4: 189473879-189473879 on Build GRCh38
rs958960623 Chr.4: 189473742-189473742 on Build GRCh38
rs564190797 Chr.4: 189473597-189473597 on Build GRCh38
rs765664178 Chr.4: 189473500-189473500 on Build GRCh38
rs151254773 Chr.4: 189473810-189473810 on Build GRCh38
rs199776124 Chr.4: 189473890-189473890 on Build GRCh38
rs773416659 Chr.4: 189473663-189473663 on Build GRCh38
rs777498576 Chr.4: 189473911-189473911 on Build GRCh38
rs998184228 Chr.4: 189473664-189473664 on Build GRCh38
rs138448031 Chr.4: 189473833-189473833 on Build GRCh38
rs200986568 Chr.4: 189473785-189473793 on Build GRCh38
rs573914859 Chr.4: 189473510-189473510 on Build GRCh38
rs138612273 Chr.4: 189473785-189473790 on Build GRCh38
rs59019208 Chr.4: 189473810-189473815 on Build GRCh38
rs149627979 Chr.4: 189473811-189473811 on Build GRCh38
rs550845911 Chr.4: 189473718-189473718 on Build GRCh38
rs918999147 Chr.4: 189473904-189473904 on Build GRCh38
rs150731190 Chr.4: 189473850-189473851 on Build GRCh38
rs1049481134 Chr.4: 189473627-189473627 on Build GRCh38
rs866929975 Chr.4: 189473807-189473807 on Build GRCh38
rs749689341 Chr.4: 189473439-189473439 on Build GRCh38
rs181527720 Chr.4: 189473728-189473728 on Build GRCh38
rs77005889 Chr.4: 189473614-189473614 on Build GRCh38
rs991573705 Chr.4: 189473754-189473754 on Build GRCh38
rs147390696 Chr.4: 189473785-189473787 on Build GRCh38
rs62340348 Chr.4: 189473565-189473565 on Build GRCh38
rs571438216 Chr.4: 189473530-189473530 on Build GRCh38
rs367983404 Chr.4: 189473889-189473889 on Build GRCh38
rs12504268 Chr.4: 189473667-189473667 on Build GRCh38
rs12511992 Chr.4: 189473918-189473918 on Build GRCh38
rs868818100 Chr.4: 189473808-189473808 on Build GRCh38
rs777817410 Chr.4: 189473788-189473793 on Build GRCh38
rs573211780 Chr.4: 189473832-189473832 on Build GRCh38
rs747116250 Chr.4: 189473934-189473934 on Build GRCh38
rs897703394 Chr.4: 189473574-189473574 on Build GRCh38
rs775489046 Chr.4: 189473481-189473481 on Build GRCh38
rs557895436 Chr.4: 189473775-189473775 on Build GRCh38
rs375941904 Chr.4: 189473807-189473815 on Build GRCh38
rs751926323 Chr.4: 189473844-189473844 on Build GRCh38
rs994667251 Chr.4: 189473590-189473590 on Build GRCh38
rs865841573 Chr.4: 189473761-189473761 on Build GRCh38
rs747875771 Chr.4: 189473432-189473432 on Build GRCh38
rs916887512 Chr.4: 189473776-189473776 on Build GRCh38
rs878892015 Chr.4: 189473788-189473790 on Build GRCh38
rs564559294 Chr.4: 189473683-189473683 on Build GRCh38
rs978886423 Chr.4: 189473686-189473686 on Build GRCh38
rs201492184 Chr.4: 189473927-189473927 on Build GRCh38
rs779063662 Chr.4: 189473446-189473446 on Build GRCh38
rs1036201452 Chr.4: 189473566-189473566 on Build GRCh38
rs889020389 Chr.4: 189473629-189473629 on Build GRCh38
rs530521367 Chr.4: 189473544-189473544 on Build GRCh38
rs762245079 Chr.4: 189473845-189473845 on Build GRCh38
rs866285250 Chr.4: 189473468-189473468 on Build GRCh38
rs544465380 Chr.4: 189473513-189473513 on Build GRCh38
rs979021585 Chr.4: 189473467-189473467 on Build GRCh38
rs765924571 Chr.4: 189473521-189473521 on Build GRCh38
rs762643451 Chr.4: 189473520-189473520 on Build GRCh38
rs560687468 Chr.4: 189473523-189473523 on Build GRCh38
rs777182009 Chr.4: 189473494-189473494 on Build GRCh38
rs930488988 Chr.4: 189473907-189473907 on Build GRCh38
rs769447625 Chr.4: 189473658-189473658 on Build GRCh38
rs555452555 Chr.4: 189473877-189473877 on Build GRCh38
rs761249257 Chr.4: 189473491-189473491 on Build GRCh38
rs185291615 Chr.4: 189473756-189473756 on Build GRCh38
rs746126511 Chr.4: 189473451-189473451 on Build GRCh38
rs113444869 Chr.4: 189473921-189473921 on Build GRCh38
rs535655536 Chr.4: 189473668-189473668 on Build GRCh38
rs752703510 Chr.4: 189473791-189473793 on Build GRCh38

View Full Product Details