Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 189475107-189475660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90AA4P
Entrez Gene ID
3323
Gene Name
heat shock protein 90 alpha family class A member 4, pseudogene
Gene Aliases
HSP90Ad, HSPCAL2
UniGene
Hs.553481
Gene Chromosome Location
Chr.4: 189472906-189475853 on Build GRCh38
Transcripts

SNP Detail

SNP ID Chromosome Location
rs758581872 Chr.4: 189475189-189475189 on Build GRCh38
rs112337873 Chr.4: 189475462-189475462 on Build GRCh38
rs997130107 Chr.4: 189475465-189475465 on Build GRCh38
rs747523719 Chr.4: 189475151-189475151 on Build GRCh38
rs772828612 Chr.4: 189475157-189475159 on Build GRCh38
rs183243005 Chr.4: 189475386-189475386 on Build GRCh38
rs1038698854 Chr.4: 189475392-189475392 on Build GRCh38
rs187545198 Chr.4: 189475535-189475535 on Build GRCh38
rs79532304 Chr.4: 189475148-189475148 on Build GRCh38
rs748572078 Chr.4: 189475159-189475159 on Build GRCh38
rs760494507 Chr.4: 189475183-189475183 on Build GRCh38
rs761630641 Chr.4: 189475495-189475495 on Build GRCh38
rs201506409 Chr.4: 189475166-189475166 on Build GRCh38
rs78036461 Chr.4: 189475575-189475575 on Build GRCh38
rs113443491 Chr.4: 189475280-189475280 on Build GRCh38
rs745894753 Chr.4: 189475458-189475458 on Build GRCh38
rs781774461 Chr.4: 189475224-189475224 on Build GRCh38
rs888047254 Chr.4: 189475352-189475352 on Build GRCh38
rs900205570 Chr.4: 189475461-189475461 on Build GRCh38
rs1047900836 Chr.4: 189475298-189475298 on Build GRCh38
rs781760680 Chr.4: 189475155-189475155 on Build GRCh38
rs528215803 Chr.4: 189475288-189475291 on Build GRCh38
rs770677538 Chr.4: 189475236-189475236 on Build GRCh38
rs116790967 Chr.4: 189475517-189475517 on Build GRCh38
rs1023546650 Chr.4: 189475569-189475569 on Build GRCh38
rs765977568 Chr.4: 189475574-189475574 on Build GRCh38
rs751745473 Chr.4: 189475214-189475214 on Build GRCh38
rs144135736 Chr.4: 189475177-189475177 on Build GRCh38
rs929482714 Chr.4: 189475295-189475295 on Build GRCh38
rs370577229 Chr.4: 189475565-189475565 on Build GRCh38
rs190797905 Chr.4: 189475607-189475607 on Build GRCh38
rs957986136 Chr.4: 189475556-189475556 on Build GRCh38
rs549960508 Chr.4: 189475187-189475187 on Build GRCh38
rs148489278 Chr.4: 189475176-189475176 on Build GRCh38
rs774226801 Chr.4: 189475490-189475490 on Build GRCh38
rs769374874 Chr.4: 189475149-189475149 on Build GRCh38
rs769352745 Chr.4: 189475154-189475154 on Build GRCh38
rs538846967 Chr.4: 189475274-189475274 on Build GRCh38
rs780125088 Chr.4: 189475147-189475147 on Build GRCh38
rs574922864 Chr.4: 189475291-189475291 on Build GRCh38
rs560173733 Chr.4: 189475163-189475163 on Build GRCh38
rs755086052 Chr.4: 189475221-189475221 on Build GRCh38
rs990177484 Chr.4: 189475179-189475179 on Build GRCh38
rs770753107 Chr.4: 189475164-189475166 on Build GRCh38
rs748623054 Chr.4: 189475225-189475225 on Build GRCh38
rs373633016 Chr.4: 189475167-189475167 on Build GRCh38
rs780133615 Chr.4: 189475199-189475199 on Build GRCh38
rs750626739 Chr.4: 189475188-189475188 on Build GRCh38
rs748866482 Chr.4: 189475180-189475180 on Build GRCh38
rs555312223 Chr.4: 189475506-189475506 on Build GRCh38
rs750619588 Chr.4: 189475134-189475134 on Build GRCh38
rs748894486 Chr.4: 189475227-189475227 on Build GRCh38
rs773973709 Chr.4: 189475238-189475238 on Build GRCh38
rs531605933 Chr.4: 189475181-189475181 on Build GRCh38
rs113840649 Chr.4: 189475136-189475136 on Build GRCh38
rs571758329 Chr.4: 189475226-189475226 on Build GRCh38
rs770358191 Chr.4: 189475172-189475172 on Build GRCh38
rs773412129 Chr.4: 189475445-189475445 on Build GRCh38
rs373342540 Chr.4: 189475384-189475385 on Build GRCh38
rs758429934 Chr.4: 189475146-189475146 on Build GRCh38

View Full Product Details