Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3024267-3024814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTATSF1P2
Entrez Gene ID
401233
Gene Name
HIV-1 Tat specific factor 1 pseudogene 2
Gene Aliases
-
UniGene
Hs.441039
Gene Chromosome Location
Chr.6: 3020156-3024771 on Build GRCh38
Transcripts
NR_033884.1 NR_132750.1

Gene Symbol
LOC101927759
Entrez Gene ID
101927759
Gene Name
uncharacterized LOC101927759
Gene Aliases
-
UniGene
Hs.145597
Gene Chromosome Location
Chr.6: 3024791-3027424 on Build GRCh38
Transcripts
NR_033884.1 NR_132750.1

SNP Detail

SNP ID Chromosome Location
rs899751411 Chr.6: 3024571-3024571 on Build GRCh38
rs62392504 Chr.6: 3024619-3024619 on Build GRCh38
rs567633679 Chr.6: 3024752-3024752 on Build GRCh38
rs909878354 Chr.6: 3024728-3024728 on Build GRCh38
rs916797905 Chr.6: 3024448-3024448 on Build GRCh38
rs957063221 Chr.6: 3024708-3024708 on Build GRCh38
rs777933822 Chr.6: 3024370-3024370 on Build GRCh38
rs912243301 Chr.6: 3024501-3024501 on Build GRCh38
rs112773018 Chr.6: 3024723-3024727 on Build GRCh38
rs540060706 Chr.6: 3024530-3024530 on Build GRCh38
rs76796586 Chr.6: 3024415-3024415 on Build GRCh38
rs960665369 Chr.6: 3024435-3024435 on Build GRCh38
rs781588174 Chr.6: 3024706-3024706 on Build GRCh38
rs991552528 Chr.6: 3024750-3024750 on Build GRCh38
rs750470518 Chr.6: 3024592-3024592 on Build GRCh38
rs1014911374 Chr.6: 3024429-3024429 on Build GRCh38
rs987910897 Chr.6: 3024778-3024778 on Build GRCh38
rs1014454041 Chr.6: 3024339-3024339 on Build GRCh38
rs34543772 Chr.6: 3024464-3024465 on Build GRCh38
rs950246901 Chr.6: 3024760-3024760 on Build GRCh38
rs1011193482 Chr.6: 3024371-3024371 on Build GRCh38
rs141306519 Chr.6: 3024328-3024328 on Build GRCh38
rs531088221 Chr.6: 3024693-3024693 on Build GRCh38
rs995338564 Chr.6: 3024588-3024588 on Build GRCh38
rs888635846 Chr.6: 3024301-3024301 on Build GRCh38
rs35794474 Chr.6: 3024352-3024352 on Build GRCh38
rs185214678 Chr.6: 3024307-3024307 on Build GRCh38
rs887885956 Chr.6: 3024601-3024601 on Build GRCh38
rs71560957 Chr.6: 3024349-3024352 on Build GRCh38
rs755323940 Chr.6: 3024689-3024689 on Build GRCh38
rs558316506 Chr.6: 3024473-3024473 on Build GRCh38
rs1024606368 Chr.6: 3024343-3024343 on Build GRCh38
rs187352112 Chr.6: 3024788-3024788 on Build GRCh38
rs538582238 Chr.6: 3024445-3024445 on Build GRCh38
rs386696228 Chr.6: 3024476-3024478 on Build GRCh38
rs983879755 Chr.6: 3024420-3024420 on Build GRCh38
rs145073170 Chr.6: 3024476-3024476 on Build GRCh38
rs943740353 Chr.6: 3024782-3024782 on Build GRCh38
rs948212092 Chr.6: 3024458-3024458 on Build GRCh38
rs62392502 Chr.6: 3024350-3024350 on Build GRCh38
rs912279313 Chr.6: 3024780-3024780 on Build GRCh38
rs1045856079 Chr.6: 3024770-3024770 on Build GRCh38
rs1052101957 Chr.6: 3024628-3024628 on Build GRCh38
rs200112588 Chr.6: 3024347-3024348 on Build GRCh38
rs4269412 Chr.6: 3024652-3024652 on Build GRCh38
rs138856583 Chr.6: 3024478-3024478 on Build GRCh38
rs1015397636 Chr.6: 3024621-3024621 on Build GRCh38
rs111462803 Chr.6: 3024685-3024685 on Build GRCh38
rs916048837 Chr.6: 3024548-3024548 on Build GRCh38
rs62392505 Chr.6: 3024684-3024684 on Build GRCh38
rs533070250 Chr.6: 3024771-3024772 on Build GRCh38
rs925461317 Chr.6: 3024694-3024694 on Build GRCh38
rs963129845 Chr.6: 3024744-3024744 on Build GRCh38
rs574658942 Chr.6: 3024523-3024523 on Build GRCh38
rs368300576 Chr.6: 3024542-3024545 on Build GRCh38
rs963963881 Chr.6: 3024421-3024421 on Build GRCh38
rs922617343 Chr.6: 3024468-3024468 on Build GRCh38
rs960701387 Chr.6: 3024642-3024642 on Build GRCh38
rs35760482 Chr.6: 3024349-3024350 on Build GRCh38
rs550832941 Chr.6: 3024712-3024712 on Build GRCh38
rs756723483 Chr.6: 3024310-3024310 on Build GRCh38
rs894761540 Chr.6: 3024338-3024338 on Build GRCh38
rs540587641 Chr.6: 3024480-3024480 on Build GRCh38
rs1056403000 Chr.6: 3024483-3024483 on Build GRCh38
rs62392501 Chr.6: 3024349-3024349 on Build GRCh38
rs1039604414 Chr.6: 3024563-3024563 on Build GRCh38

View Full Product Details