Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145895750-145895608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
UniGene
Hs.158237
Gene Chromosome Location
Chr.1: 145891208-145910189 on Build GRCh38
Transcripts
NM_001303040.1 NM_001303041.1 NM_003637.4 XM_005277436.3 XM_011510083.2 XM_011510084.1 XM_017002622.1 XM_017002623.1 XM_017002624.1 XM_017002625.1 XM_017002626.1 XM_017002628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
895195 c.3060T>C p.T1020T Chr.1: 145895685-145895685 on Build GRCh38
1667889 c.3095T>G p.L1032R Chr.1: 145895650-145895650 on Build GRCh38
6233253 c.3043A>G p.I1015V Chr.1: 145895702-145895702 on Build GRCh38
6687771 c.3065C>T p.P1022L Chr.1: 145895680-145895680 on Build GRCh38
4541889 c.3088G>A p.E1030K Chr.1: 145895657-145895657 on Build GRCh38
View More

View Full Product Details