Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159991192-159991358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NM_000876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741325 c.271C>G p.R91G Chr.6: 159991305-159991305 on Build GRCh38
5609113 c.288G>T p.V96V Chr.6: 159991322-159991322 on Build GRCh38
6201973 c.234G>A p.G78G Chr.6: 159991268-159991268 on Build GRCh38
1441945 c.272G>A p.R91H Chr.6: 159991306-159991306 on Build GRCh38
3023451 c.249T>G p.V83V Chr.6: 159991283-159991283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150881942 Chr.6: 159991313-159991313 on Build GRCh38
rs143824089 Chr.6: 159991308-159991308 on Build GRCh38
rs775879753 Chr.6: 159991259-159991259 on Build GRCh38
rs763714994 Chr.6: 159991297-159991297 on Build GRCh38
rs752296293 Chr.6: 159991237-159991237 on Build GRCh38
rs147110717 Chr.6: 159991281-159991281 on Build GRCh38
rs201467924 Chr.6: 159991258-159991258 on Build GRCh38
rs767878929 Chr.6: 159991271-159991271 on Build GRCh38
rs773726244 Chr.6: 159991274-159991274 on Build GRCh38
rs199950272 Chr.6: 159991305-159991305 on Build GRCh38
rs766465285 Chr.6: 159991230-159991230 on Build GRCh38
rs751957182 Chr.6: 159991263-159991264 on Build GRCh38
rs1015964251 Chr.6: 159991309-159991309 on Build GRCh38
rs747031754 Chr.6: 159991246-159991246 on Build GRCh38
rs757915126 Chr.6: 159991243-159991243 on Build GRCh38
rs372323465 Chr.6: 159991262-159991262 on Build GRCh38
rs140560792 Chr.6: 159991265-159991265 on Build GRCh38
rs201786202 Chr.6: 159991330-159991330 on Build GRCh38
rs762430159 Chr.6: 159991266-159991266 on Build GRCh38
rs148344348 Chr.6: 159991236-159991236 on Build GRCh38
rs8191704 Chr.6: 159991306-159991306 on Build GRCh38
rs768749227 Chr.6: 159991315-159991315 on Build GRCh38
rs769674629 Chr.6: 159991257-159991257 on Build GRCh38
rs752349284 Chr.6: 159991292-159991292 on Build GRCh38
rs750730242 Chr.6: 159991234-159991234 on Build GRCh38
rs758037852 Chr.6: 159991293-159991293 on Build GRCh38
rs781093873 Chr.6: 159991254-159991254 on Build GRCh38
rs745629855 Chr.6: 159991255-159991255 on Build GRCh38
rs138574174 Chr.6: 159991298-159991298 on Build GRCh38
rs757132015 Chr.6: 159991252-159991252 on Build GRCh38
rs766668837 Chr.6: 159991288-159991288 on Build GRCh38
rs749466130 Chr.6: 159991261-159991261 on Build GRCh38

View Full Product Details