Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42925461-42925631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK3
Entrez Gene ID
84376
Gene Name
hook microtubule-tethering protein 3
Gene Aliases
HK3
UniGene
Hs.162852
Gene Chromosome Location
Chr.8: 42896890-43030539 on Build GRCh38
Transcripts
NM_032410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7119932 c.148C>G p.P50A Chr.8: 42925561-42925561 on Build GRCh38
2962070 c.145G>T p.D49Y Chr.8: 42925558-42925558 on Build GRCh38
4854136 c.148C>A p.P50T Chr.8: 42925561-42925561 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779561470 Chr.8: 42925590-42925590 on Build GRCh38
rs201428376 Chr.8: 42925567-42925567 on Build GRCh38
rs745378155 Chr.8: 42925542-42925545 on Build GRCh38
rs765668561 Chr.8: 42925526-42925526 on Build GRCh38
rs773594780 Chr.8: 42925515-42925515 on Build GRCh38
rs763289021 Chr.8: 42925530-42925530 on Build GRCh38
rs936801643 Chr.8: 42925574-42925574 on Build GRCh38
rs370980704 Chr.8: 42925524-42925524 on Build GRCh38
rs375541096 Chr.8: 42925536-42925536 on Build GRCh38
rs781352180 Chr.8: 42925553-42925553 on Build GRCh38
rs867142327 Chr.8: 42925564-42925564 on Build GRCh38
rs755118749 Chr.8: 42925548-42925548 on Build GRCh38
rs897033675 Chr.8: 42925541-42925541 on Build GRCh38
rs533962099 Chr.8: 42925594-42925594 on Build GRCh38
rs918188524 Chr.8: 42925521-42925521 on Build GRCh38
rs762244200 Chr.8: 42925517-42925517 on Build GRCh38
rs757092870 Chr.8: 42925554-42925554 on Build GRCh38
rs140974160 Chr.8: 42925581-42925581 on Build GRCh38
rs557883506 Chr.8: 42925566-42925566 on Build GRCh38

View Full Product Details