Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160010573-160010736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
UniGene
Hs.487062
Gene Chromosome Location
Chr.6: 159969099-160111504 on Build GRCh38
Transcripts
NM_000876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3921017 c.421C>T p.P141S Chr.6: 160010693-160010693 on Build GRCh38
1311785 c.421C>A p.P141T Chr.6: 160010693-160010693 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758656646 Chr.6: 160010707-160010707 on Build GRCh38
rs187597600 Chr.6: 160010690-160010690 on Build GRCh38
rs761433287 Chr.6: 160010658-160010658 on Build GRCh38
rs772958120 Chr.6: 160010663-160010663 on Build GRCh38
rs760282651 Chr.6: 160010671-160010671 on Build GRCh38
rs764995243 Chr.6: 160010691-160010691 on Build GRCh38
rs369524729 Chr.6: 160010682-160010682 on Build GRCh38
rs758024193 Chr.6: 160010636-160010636 on Build GRCh38
rs963229795 Chr.6: 160010655-160010655 on Build GRCh38
rs973635025 Chr.6: 160010604-160010604 on Build GRCh38
rs759510003 Chr.6: 160010688-160010688 on Build GRCh38
rs929376664 Chr.6: 160010633-160010633 on Build GRCh38
rs919056332 Chr.6: 160010621-160010621 on Build GRCh38
rs765929352 Chr.6: 160010673-160010673 on Build GRCh38
rs199536051 Chr.6: 160010656-160010656 on Build GRCh38
rs752512454 Chr.6: 160010695-160010695 on Build GRCh38
rs200930371 Chr.6: 160010660-160010660 on Build GRCh38
rs3215571 Chr.6: 160010672-160010673 on Build GRCh38
rs777377266 Chr.6: 160010656-160010656 on Build GRCh38
rs559945419 Chr.6: 160010603-160010603 on Build GRCh38
rs8191741 Chr.6: 160010681-160010682 on Build GRCh38
rs534646403 Chr.6: 160010599-160010599 on Build GRCh38
rs372596601 Chr.6: 160010634-160010634 on Build GRCh38
rs770815621 Chr.6: 160010673-160010673 on Build GRCh38
rs770059719 Chr.6: 160010649-160010649 on Build GRCh38

View Full Product Details