Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35873526-35873676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
UniGene
Hs.591742
Gene Chromosome Location
Chr.5: 35856875-35879603 on Build GRCh38
Transcripts
NM_002185.3 NR_120485.1 XM_005248299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
233970 c.706+6G>A p.? Chr.5: 35873654-35873654 on Build GRCh38
5403379 c.586C>T p.Q196* Chr.5: 35873528-35873528 on Build GRCh38
7342389 c.602A>G p.Y201C Chr.5: 35873544-35873544 on Build GRCh38
6921337 c.634T>A p.Y212N Chr.5: 35873576-35873576 on Build GRCh38
1254793 c.667A>G p.S223G Chr.5: 35873609-35873609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772747002 Chr.5: 35873572-35873572 on Build GRCh38
rs786205572 Chr.5: 35873578-35873578 on Build GRCh38
rs753994938 Chr.5: 35873647-35873647 on Build GRCh38
rs901925052 Chr.5: 35873607-35873607 on Build GRCh38
rs199714011 Chr.5: 35873637-35873637 on Build GRCh38
rs761704139 Chr.5: 35873634-35873634 on Build GRCh38
rs779479151 Chr.5: 35873562-35873562 on Build GRCh38
rs997580184 Chr.5: 35873616-35873616 on Build GRCh38
rs104893893 Chr.5: 35873593-35873593 on Build GRCh38
rs774177784 Chr.5: 35873627-35873627 on Build GRCh38
rs755300062 Chr.5: 35873560-35873560 on Build GRCh38
rs148165103 Chr.5: 35873622-35873622 on Build GRCh38
rs566889429 Chr.5: 35873641-35873641 on Build GRCh38
rs193922644 Chr.5: 35873559-35873559 on Build GRCh38
rs748643880 Chr.5: 35873567-35873567 on Build GRCh38
rs769433714 Chr.5: 35873608-35873608 on Build GRCh38
rs757795302 Chr.5: 35873571-35873571 on Build GRCh38
rs587778405 Chr.5: 35873604-35873604 on Build GRCh38
rs866344764 Chr.5: 35873563-35873563 on Build GRCh38
rs201559094 Chr.5: 35873558-35873558 on Build GRCh38
rs374232919 Chr.5: 35873595-35873595 on Build GRCh38
rs193922645 Chr.5: 35873586-35873586 on Build GRCh38
rs200388101 Chr.5: 35873545-35873545 on Build GRCh38
rs762736419 Chr.5: 35873624-35873624 on Build GRCh38
rs201897242 Chr.5: 35873644-35873644 on Build GRCh38
rs766555082 Chr.5: 35873646-35873646 on Build GRCh38
rs201412341 Chr.5: 35873625-35873625 on Build GRCh38
rs1039419839 Chr.5: 35873550-35873550 on Build GRCh38
rs867618621 Chr.5: 35873546-35873546 on Build GRCh38
rs200894429 Chr.5: 35873643-35873643 on Build GRCh38
rs577344998 Chr.5: 35873577-35873577 on Build GRCh38
rs193922643 Chr.5: 35873558-35873559 on Build GRCh38

View Full Product Details