Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122729536-122729680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR1
Entrez Gene ID
27198
Gene Name
hydroxycarboxylic acid receptor 1
Gene Aliases
FKSG80, GPR104, GPR81, HCA1, LACR1, TA-GPCR, TAGPCR
UniGene
Hs.610873
Gene Chromosome Location
Chr.12: 122727606-122730582 on Build GRCh38
Transcripts
NM_032554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346335 c.804C>A p.Y268* Chr.12: 122729536-122729536 on Build GRCh38
3368701 c.742C>T p.P248S Chr.12: 122729598-122729598 on Build GRCh38
936310 c.687C>A p.F229L Chr.12: 122729653-122729653 on Build GRCh38
6249343 c.696C>A p.C232* Chr.12: 122729644-122729644 on Build GRCh38
6286312 c.783C>A p.H261Q Chr.12: 122729557-122729557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377541263 Chr.12: 122729620-122729620 on Build GRCh38
rs146760413 Chr.12: 122729584-122729584 on Build GRCh38
rs149768068 Chr.12: 122729632-122729632 on Build GRCh38
rs547953029 Chr.12: 122729603-122729603 on Build GRCh38
rs756991659 Chr.12: 122729595-122729595 on Build GRCh38
rs373609922 Chr.12: 122729602-122729602 on Build GRCh38
rs138206647 Chr.12: 122729643-122729643 on Build GRCh38
rs558552970 Chr.12: 122729642-122729642 on Build GRCh38
rs375870381 Chr.12: 122729636-122729636 on Build GRCh38
rs767771972 Chr.12: 122729567-122729567 on Build GRCh38
rs761061194 Chr.12: 122729652-122729652 on Build GRCh38
rs373136345 Chr.12: 122729591-122729591 on Build GRCh38
rs36124671 Chr.12: 122729583-122729583 on Build GRCh38
rs773312705 Chr.12: 122729647-122729647 on Build GRCh38
rs779198312 Chr.12: 122729628-122729628 on Build GRCh38
rs951420252 Chr.12: 122729594-122729594 on Build GRCh38
rs140482291 Chr.12: 122729619-122729619 on Build GRCh38
rs371072988 Chr.12: 122729631-122729631 on Build GRCh38
rs766752596 Chr.12: 122729656-122729656 on Build GRCh38
rs998780278 Chr.12: 122729585-122729585 on Build GRCh38
rs141008238 Chr.12: 122729570-122729570 on Build GRCh38
rs751207967 Chr.12: 122729590-122729590 on Build GRCh38
rs536097747 Chr.12: 122729574-122729574 on Build GRCh38
rs915639443 Chr.12: 122729565-122729565 on Build GRCh38
rs370452174 Chr.12: 122729599-122729599 on Build GRCh38
rs369713712 Chr.12: 122729660-122729660 on Build GRCh38

View Full Product Details