Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35867247-35867416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
UniGene
Hs.591742
Gene Chromosome Location
Chr.5: 35856875-35879603 on Build GRCh38
Transcripts
NM_002185.3 NR_120485.1 XM_005248299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6920161 c.262C>A p.L88I Chr.5: 35867346-35867346 on Build GRCh38
5403377 c.303G>A p.L101L Chr.5: 35867387-35867387 on Build GRCh38
1437172 c.270G>T p.E90D Chr.5: 35867354-35867354 on Build GRCh38
737979 c.282C>T p.I94I Chr.5: 35867366-35867366 on Build GRCh38
1067539 c.319A>T p.I107L Chr.5: 35867403-35867403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757797163 Chr.5: 35867329-35867329 on Build GRCh38
rs201767916 Chr.5: 35867330-35867330 on Build GRCh38
rs755030275 Chr.5: 35867311-35867311 on Build GRCh38
rs753909264 Chr.5: 35867297-35867297 on Build GRCh38
rs141698985 Chr.5: 35867349-35867349 on Build GRCh38
rs202194801 Chr.5: 35867284-35867284 on Build GRCh38
rs751425932 Chr.5: 35867383-35867385 on Build GRCh38
rs1020619032 Chr.5: 35867341-35867341 on Build GRCh38
rs552630274 Chr.5: 35867321-35867321 on Build GRCh38
rs773973629 Chr.5: 35867374-35867374 on Build GRCh38
rs910624460 Chr.5: 35867277-35867277 on Build GRCh38
rs200475138 Chr.5: 35867323-35867323 on Build GRCh38
rs193922640 Chr.5: 35867364-35867365 on Build GRCh38
rs771757728 Chr.5: 35867384-35867384 on Build GRCh38
rs773047299 Chr.5: 35867271-35867271 on Build GRCh38
rs929178646 Chr.5: 35867380-35867380 on Build GRCh38
rs200687605 Chr.5: 35867316-35867316 on Build GRCh38
rs758304802 Chr.5: 35867283-35867284 on Build GRCh38
rs760580267 Chr.5: 35867282-35867282 on Build GRCh38
rs191585195 Chr.5: 35867367-35867367 on Build GRCh38
rs139835773 Chr.5: 35867286-35867286 on Build GRCh38
rs778067695 Chr.5: 35867366-35867366 on Build GRCh38
rs766399441 Chr.5: 35867314-35867314 on Build GRCh38
rs772953676 Chr.5: 35867385-35867385 on Build GRCh38
rs747417170 Chr.5: 35867327-35867327 on Build GRCh38

View Full Product Details