Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34327767-34327994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1 XM_017027526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
710354 c.874G>T p.G292* Chr.19: 34327798-34327798 on Build GRCh38
321168 c.847-1G>C p.? Chr.19: 34327770-34327770 on Build GRCh38
6150558 c.849A>T p.A283A Chr.19: 34327773-34327773 on Build GRCh38
994745 c.923C>T p.A308V Chr.19: 34327847-34327847 on Build GRCh38
4752607 c.898G>A p.A300T Chr.19: 34327822-34327822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974388362 Chr.19: 34327965-34327965 on Build GRCh38
rs563035641 Chr.19: 34327943-34327943 on Build GRCh38
rs756258708 Chr.19: 34327870-34327870 on Build GRCh38
rs773745783 Chr.19: 34327950-34327951 on Build GRCh38
rs201554641 Chr.19: 34327815-34327815 on Build GRCh38
rs56236221 Chr.19: 34327911-34327911 on Build GRCh38
rs59405414 Chr.19: 34327927-34327927 on Build GRCh38
rs542445298 Chr.19: 34327962-34327962 on Build GRCh38
rs923535045 Chr.19: 34327968-34327968 on Build GRCh38
rs535714834 Chr.19: 34327793-34327793 on Build GRCh38
rs199757501 Chr.19: 34327951-34327951 on Build GRCh38
rs369591873 Chr.19: 34327949-34327949 on Build GRCh38
rs972984761 Chr.19: 34327910-34327910 on Build GRCh38
rs952847606 Chr.19: 34327912-34327912 on Build GRCh38
rs200928748 Chr.19: 34327824-34327824 on Build GRCh38
rs755516681 Chr.19: 34327884-34327884 on Build GRCh38
rs60232252 Chr.19: 34327803-34327803 on Build GRCh38
rs762079775 Chr.19: 34327842-34327842 on Build GRCh38
rs752539191 Chr.19: 34327836-34327836 on Build GRCh38
rs748566294 Chr.19: 34327900-34327900 on Build GRCh38
rs943044743 Chr.19: 34327933-34327933 on Build GRCh38
rs768142103 Chr.19: 34327892-34327893 on Build GRCh38
rs762216763 Chr.19: 34327958-34327958 on Build GRCh38
rs576582597 Chr.19: 34327950-34327950 on Build GRCh38
rs770414673 Chr.19: 34327913-34327913 on Build GRCh38
rs368749949 Chr.19: 34327827-34327827 on Build GRCh38
rs777370414 Chr.19: 34327905-34327905 on Build GRCh38
rs781647625 Chr.19: 34327901-34327901 on Build GRCh38
rs771739573 Chr.19: 34327806-34327806 on Build GRCh38
rs754244135 Chr.19: 34327878-34327878 on Build GRCh38
rs757412778 Chr.19: 34327847-34327847 on Build GRCh38
rs770464094 Chr.19: 34327825-34327825 on Build GRCh38
rs780342573 Chr.19: 34327871-34327871 on Build GRCh38
rs776426974 Chr.19: 34327826-34327826 on Build GRCh38
rs779104776 Chr.19: 34327893-34327893 on Build GRCh38
rs144093159 Chr.19: 34327833-34327833 on Build GRCh38
rs772470083 Chr.19: 34327904-34327904 on Build GRCh38
rs199991533 Chr.19: 34327848-34327848 on Build GRCh38

View Full Product Details