Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78173710-78173959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD21
Entrez Gene ID
283219
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
KCASH2
UniGene
Hs.528439
Gene Chromosome Location
Chr.11: 78171243-78188854 on Build GRCh38
Transcripts
NM_001029859.2 XM_005273925.3 XM_005273928.1 XM_006718516.3 XM_006718517.2 XM_006718518.3 XM_011544955.2 NR_102280.1 NR_102281.1

Gene Symbol
KCTD21-AS1
Entrez Gene ID
100289388
Gene Name
KCTD21 antisense RNA 1
Gene Aliases
-
UniGene
Hs.628183
Gene Chromosome Location
Chr.11: 78139793-78173977 on Build GRCh38
Transcripts
NM_001029859.2 XM_005273925.3 XM_005273928.1 XM_006718516.3 XM_006718517.2 XM_006718518.3 XM_011544955.2 NR_102280.1 NR_102281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036904 c.704G>A p.G235D Chr.11: 78173851-78173851 on Build GRCh38
4036903 c.752A>G p.N251S Chr.11: 78173803-78173803 on Build GRCh38
2019251 c.733G>A p.A245T Chr.11: 78173822-78173822 on Build GRCh38
7096222 c.774delG p.Y259fs*>2 Chr.11: 78173781-78173781 on Build GRCh38
167356 c.763C>T p.R255* Chr.11: 78173792-78173792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982628862 Chr.11: 78173829-78173829 on Build GRCh38
rs923874401 Chr.11: 78173909-78173909 on Build GRCh38
rs972004623 Chr.11: 78173793-78173793 on Build GRCh38
rs761167201 Chr.11: 78173759-78173759 on Build GRCh38
rs766947707 Chr.11: 78173834-78173852 on Build GRCh38
rs769308293 Chr.11: 78173734-78173734 on Build GRCh38
rs200360563 Chr.11: 78173834-78173834 on Build GRCh38
rs373812384 Chr.11: 78173885-78173885 on Build GRCh38
rs372134817 Chr.11: 78173791-78173791 on Build GRCh38
rs146120097 Chr.11: 78173841-78173841 on Build GRCh38
rs765435164 Chr.11: 78173768-78173768 on Build GRCh38
rs370351340 Chr.11: 78173860-78173860 on Build GRCh38
rs764015451 Chr.11: 78173877-78173877 on Build GRCh38
rs1042367767 Chr.11: 78173902-78173902 on Build GRCh38
rs758533741 Chr.11: 78173776-78173776 on Build GRCh38
rs755490315 Chr.11: 78173766-78173766 on Build GRCh38
rs201151831 Chr.11: 78173919-78173919 on Build GRCh38
rs766583173 Chr.11: 78173761-78173761 on Build GRCh38
rs748855021 Chr.11: 78173840-78173840 on Build GRCh38
rs762418147 Chr.11: 78173740-78173740 on Build GRCh38
rs368809886 Chr.11: 78173746-78173746 on Build GRCh38
rs747863144 Chr.11: 78173928-78173928 on Build GRCh38
rs755890166 Chr.11: 78173915-78173915 on Build GRCh38
rs373870024 Chr.11: 78173729-78173729 on Build GRCh38
rs972959546 Chr.11: 78173755-78173755 on Build GRCh38
rs867538587 Chr.11: 78173739-78173739 on Build GRCh38
rs759951749 Chr.11: 78173858-78173858 on Build GRCh38
rs752867312 Chr.11: 78173769-78173769 on Build GRCh38
rs780030980 Chr.11: 78173917-78173917 on Build GRCh38
rs745828501 Chr.11: 78173826-78173826 on Build GRCh38
rs202161857 Chr.11: 78173855-78173855 on Build GRCh38
rs757080401 Chr.11: 78173792-78173792 on Build GRCh38
rs778118017 Chr.11: 78173782-78173782 on Build GRCh38
rs138655552 Chr.11: 78173843-78173843 on Build GRCh38
rs759404638 Chr.11: 78173732-78173732 on Build GRCh38
rs777091754 Chr.11: 78173857-78173857 on Build GRCh38
rs201143591 Chr.11: 78173783-78173783 on Build GRCh38
rs753245272 Chr.11: 78173864-78173864 on Build GRCh38
rs780794697 Chr.11: 78173805-78173805 on Build GRCh38
rs186087116 Chr.11: 78173906-78173906 on Build GRCh38
rs757372273 Chr.11: 78173886-78173886 on Build GRCh38
rs181912239 Chr.11: 78173810-78173810 on Build GRCh38
rs769679625 Chr.11: 78173828-78173828 on Build GRCh38
rs230661 Chr.11: 78173736-78173736 on Build GRCh38
rs773860364 Chr.11: 78173758-78173758 on Build GRCh38
rs377058404 Chr.11: 78173856-78173856 on Build GRCh38
rs754380540 Chr.11: 78173764-78173764 on Build GRCh38
rs755258463 Chr.11: 78173744-78173744 on Build GRCh38
rs763785078 Chr.11: 78173754-78173754 on Build GRCh38
rs80062567 Chr.11: 78173865-78173865 on Build GRCh38
rs749154861 Chr.11: 78173918-78173918 on Build GRCh38
rs773682782 Chr.11: 78173922-78173922 on Build GRCh38

View Full Product Details