Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7726899-7727172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
UniGene
Hs.75285
Gene Chromosome Location
Chr.10: 7703273-7749520 on Build GRCh38
Transcripts
NM_002216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190670 c.1063A>G p.I355V Chr.10: 7727028-7727028 on Build GRCh38
920582 c.1143G>A p.Q381Q Chr.10: 7727108-7727108 on Build GRCh38
3440716 c.1073G>A p.W358* Chr.10: 7727038-7727038 on Build GRCh38
3867858 c.1093G>A p.A365T Chr.10: 7727058-7727058 on Build GRCh38
920581 c.1107G>T p.Q369H Chr.10: 7727072-7727072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563195255 Chr.10: 7727000-7727000 on Build GRCh38
rs778746391 Chr.10: 7727120-7727120 on Build GRCh38
rs545967088 Chr.10: 7727031-7727031 on Build GRCh38
rs201131091 Chr.10: 7726951-7726951 on Build GRCh38
rs371925713 Chr.10: 7726936-7726936 on Build GRCh38
rs990145199 Chr.10: 7726947-7726947 on Build GRCh38
rs1003691864 Chr.10: 7727029-7727029 on Build GRCh38
rs763879060 Chr.10: 7726983-7726983 on Build GRCh38
rs374860908 Chr.10: 7726972-7726972 on Build GRCh38
rs200104158 Chr.10: 7726970-7726970 on Build GRCh38
rs559797350 Chr.10: 7727095-7727095 on Build GRCh38
rs771294645 Chr.10: 7727038-7727038 on Build GRCh38
rs752371503 Chr.10: 7727123-7727123 on Build GRCh38
rs201049656 Chr.10: 7727081-7727081 on Build GRCh38
rs777189918 Chr.10: 7726952-7726952 on Build GRCh38
rs781598953 Chr.10: 7727039-7727039 on Build GRCh38
rs777394781 Chr.10: 7727082-7727083 on Build GRCh38
rs118030654 Chr.10: 7727080-7727080 on Build GRCh38
rs889474561 Chr.10: 7727113-7727113 on Build GRCh38
rs763174219 Chr.10: 7727060-7727060 on Build GRCh38
rs750359604 Chr.10: 7727090-7727090 on Build GRCh38
rs1037269224 Chr.10: 7727058-7727058 on Build GRCh38
rs768936398 Chr.10: 7727071-7727071 on Build GRCh38
rs760594995 Chr.10: 7727091-7727091 on Build GRCh38
rs2274455 Chr.10: 7727138-7727138 on Build GRCh38
rs747566791 Chr.10: 7726940-7726940 on Build GRCh38
rs185942404 Chr.10: 7727032-7727032 on Build GRCh38
rs143417500 Chr.10: 7727022-7727022 on Build GRCh38
rs532614781 Chr.10: 7727046-7727046 on Build GRCh38
rs774528846 Chr.10: 7727078-7727078 on Build GRCh38
rs368254410 Chr.10: 7726924-7726924 on Build GRCh38
rs541730267 Chr.10: 7726990-7726990 on Build GRCh38
rs769978719 Chr.10: 7726965-7726973 on Build GRCh38
rs983357930 Chr.10: 7727135-7727135 on Build GRCh38
rs755605147 Chr.10: 7727013-7727013 on Build GRCh38
rs754529002 Chr.10: 7727114-7727114 on Build GRCh38
rs552493791 Chr.10: 7726929-7726929 on Build GRCh38
rs146728457 Chr.10: 7727005-7727005 on Build GRCh38
rs759574095 Chr.10: 7726969-7726969 on Build GRCh38
rs763117322 Chr.10: 7726973-7726973 on Build GRCh38
rs753459183 Chr.10: 7727108-7727108 on Build GRCh38
rs771398424 Chr.10: 7726942-7726942 on Build GRCh38
rs772595952 Chr.10: 7726934-7726934 on Build GRCh38
rs753545358 Chr.10: 7727018-7727018 on Build GRCh38
rs779671708 Chr.10: 7727017-7727017 on Build GRCh38
rs775477808 Chr.10: 7727056-7727056 on Build GRCh38
rs138958445 Chr.10: 7727045-7727045 on Build GRCh38

View Full Product Details