Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119047728-119047995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100761 c.2475C>T p.L825L Chr.11: 119047982-119047982 on Build GRCh38
5837970 c.2503T>C p.F835L Chr.11: 119047954-119047954 on Build GRCh38
4760599 c.2572G>C p.E858Q Chr.11: 119047757-119047757 on Build GRCh38
7177486 c.2492A>G p.H831R Chr.11: 119047965-119047965 on Build GRCh38
923712 c.2506C>T p.L836F Chr.11: 119047951-119047951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373632853 Chr.11: 119047911-119047911 on Build GRCh38
rs151232109 Chr.11: 119047952-119047952 on Build GRCh38
rs781917127 Chr.11: 119047810-119047810 on Build GRCh38
rs899115152 Chr.11: 119047828-119047828 on Build GRCh38
rs782785661 Chr.11: 119047961-119047961 on Build GRCh38
rs782591504 Chr.11: 119047909-119047909 on Build GRCh38
rs145651462 Chr.11: 119047863-119047863 on Build GRCh38
rs371646359 Chr.11: 119047754-119047754 on Build GRCh38
rs782257624 Chr.11: 119047762-119047762 on Build GRCh38
rs782712060 Chr.11: 119047947-119047947 on Build GRCh38
rs782174368 Chr.11: 119047913-119047913 on Build GRCh38
rs781969909 Chr.11: 119047941-119047941 on Build GRCh38
rs782166950 Chr.11: 119047957-119047957 on Build GRCh38
rs2298481 Chr.11: 119047937-119047937 on Build GRCh38
rs782627824 Chr.11: 119047848-119047848 on Build GRCh38
rs782014723 Chr.11: 119047921-119047921 on Build GRCh38
rs544870491 Chr.11: 119047834-119047834 on Build GRCh38
rs782206626 Chr.11: 119047793-119047793 on Build GRCh38
rs187600980 Chr.11: 119047773-119047773 on Build GRCh38
rs1047618126 Chr.11: 119047870-119047870 on Build GRCh38
rs782470212 Chr.11: 119047906-119047906 on Build GRCh38
rs61902088 Chr.11: 119047811-119047811 on Build GRCh38
rs138286788 Chr.11: 119047970-119047970 on Build GRCh38
rs782089925 Chr.11: 119047944-119047944 on Build GRCh38
rs1054653867 Chr.11: 119047839-119047839 on Build GRCh38
rs912190885 Chr.11: 119047761-119047761 on Build GRCh38
rs376544274 Chr.11: 119047871-119047871 on Build GRCh38
rs575171212 Chr.11: 119047919-119047919 on Build GRCh38
rs374558947 Chr.11: 119047814-119047814 on Build GRCh38
rs782264496 Chr.11: 119047924-119047924 on Build GRCh38
rs782319880 Chr.11: 119047797-119047797 on Build GRCh38
rs372174234 Chr.11: 119047813-119047813 on Build GRCh38
rs1008700021 Chr.11: 119047873-119047873 on Build GRCh38
rs7931970 Chr.11: 119047825-119047825 on Build GRCh38
rs782370481 Chr.11: 119047765-119047765 on Build GRCh38
rs781822973 Chr.11: 119047762-119047764 on Build GRCh38
rs782105820 Chr.11: 119047761-119047762 on Build GRCh38
rs782780774 Chr.11: 119047766-119047766 on Build GRCh38
rs1019746764 Chr.11: 119047902-119047902 on Build GRCh38
rs782697970 Chr.11: 119047917-119047917 on Build GRCh38
rs782514771 Chr.11: 119047845-119047845 on Build GRCh38
rs201472881 Chr.11: 119047818-119047818 on Build GRCh38
rs781880887 Chr.11: 119047842-119047842 on Build GRCh38
rs782764646 Chr.11: 119047970-119047972 on Build GRCh38
rs782796557 Chr.11: 119047812-119047812 on Build GRCh38

View Full Product Details