Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151437567-151437826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF10
Entrez Gene ID
285313
Gene Name
immunoglobulin superfamily member 10
Gene Aliases
CMF608
UniGene
Hs.58561
Gene Chromosome Location
Chr.3: 151435051-151619926 on Build GRCh38
Transcripts
NM_001178145.1 NM_001178146.1 NM_178822.4 XM_011512708.2 XM_011512709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163785 c.6841C>T p.L2281F Chr.3: 151437720-151437720 on Build GRCh38
206521 c.6967G>A p.V2323M Chr.3: 151437594-151437594 on Build GRCh38
4692011 c.6771delA p.K2257fs*47 Chr.3: 151437790-151437790 on Build GRCh38
6684498 c.6946C>T p.R2316* Chr.3: 151437615-151437615 on Build GRCh38
4114498 c.6801A>G p.P2267P Chr.3: 151437760-151437760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777232976 Chr.3: 151437695-151437706 on Build GRCh38
rs764206051 Chr.3: 151437607-151437607 on Build GRCh38
rs763240944 Chr.3: 151437748-151437749 on Build GRCh38
rs201769254 Chr.3: 151437625-151437625 on Build GRCh38
rs766036065 Chr.3: 151437779-151437780 on Build GRCh38
rs780539145 Chr.3: 151437790-151437790 on Build GRCh38
rs751857689 Chr.3: 151437630-151437630 on Build GRCh38
rs762281347 Chr.3: 151437723-151437724 on Build GRCh38
rs765451327 Chr.3: 151437767-151437767 on Build GRCh38
rs375058796 Chr.3: 151437697-151437697 on Build GRCh38
rs761020196 Chr.3: 151437605-151437605 on Build GRCh38
rs529047696 Chr.3: 151437642-151437642 on Build GRCh38
rs763865273 Chr.3: 151437681-151437681 on Build GRCh38
rs764073002 Chr.3: 151437624-151437627 on Build GRCh38
rs751019784 Chr.3: 151437615-151437615 on Build GRCh38
rs150554446 Chr.3: 151437731-151437731 on Build GRCh38
rs762106625 Chr.3: 151437765-151437765 on Build GRCh38
rs761986013 Chr.3: 151437613-151437613 on Build GRCh38
rs751161807 Chr.3: 151437784-151437785 on Build GRCh38
rs1016525929 Chr.3: 151437796-151437796 on Build GRCh38
rs754584576 Chr.3: 151437789-151437790 on Build GRCh38
rs754052654 Chr.3: 151437608-151437608 on Build GRCh38
rs774302298 Chr.3: 151437594-151437594 on Build GRCh38
rs368203195 Chr.3: 151437701-151437701 on Build GRCh38
rs201962174 Chr.3: 151437634-151437634 on Build GRCh38
rs750821189 Chr.3: 151437737-151437739 on Build GRCh38
rs1005434748 Chr.3: 151437757-151437758 on Build GRCh38
rs780539580 Chr.3: 151437628-151437628 on Build GRCh38
rs765361967 Chr.3: 151437614-151437614 on Build GRCh38
rs900503520 Chr.3: 151437640-151437640 on Build GRCh38
rs41401648 Chr.3: 151437595-151437595 on Build GRCh38
rs765680065 Chr.3: 151437728-151437729 on Build GRCh38
rs763573934 Chr.3: 151437799-151437799 on Build GRCh38
rs768428095 Chr.3: 151437737-151437737 on Build GRCh38
rs891202112 Chr.3: 151437789-151437789 on Build GRCh38
rs997640867 Chr.3: 151437644-151437644 on Build GRCh38
rs371755582 Chr.3: 151437596-151437596 on Build GRCh38
rs369523098 Chr.3: 151437751-151437751 on Build GRCh38
rs750530453 Chr.3: 151437774-151437774 on Build GRCh38
rs371260847 Chr.3: 151437683-151437683 on Build GRCh38
rs199576775 Chr.3: 151437622-151437622 on Build GRCh38
rs777297326 Chr.3: 151437633-151437633 on Build GRCh38

View Full Product Details