Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 125043778-125044021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0196
Entrez Gene ID
9897
Gene Name
KIAA0196
Gene Aliases
RTSC, RTSC1, SPG8
UniGene
Hs.270043
Gene Chromosome Location
Chr.8: 125024260-125091819 on Build GRCh38
Transcripts
NM_014846.3 XM_005251120.3 XM_011517409.1 XM_017014113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964079 c.2794T>C p.S932P Chr.8: 125043881-125043881 on Build GRCh38
2151427 c.2797G>A p.A933T Chr.8: 125043878-125043878 on Build GRCh38
5873895 c.2793T>A p.F931L Chr.8: 125043882-125043882 on Build GRCh38
1095957 c.2746G>A p.V916I Chr.8: 125044016-125044016 on Build GRCh38
3896674 c.2828C>T p.A943V Chr.8: 125043847-125043847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759801315 Chr.8: 125043833-125043833 on Build GRCh38
rs181946889 Chr.8: 125043906-125043906 on Build GRCh38
rs67806006 Chr.8: 125043974-125043975 on Build GRCh38
rs200770153 Chr.8: 125043875-125043875 on Build GRCh38
rs746751439 Chr.8: 125043820-125043820 on Build GRCh38
rs767992061 Chr.8: 125043976-125043976 on Build GRCh38
rs1024430224 Chr.8: 125043836-125043836 on Build GRCh38
rs11370883 Chr.8: 125043973-125043974 on Build GRCh38
rs952413031 Chr.8: 125043856-125043856 on Build GRCh38
rs749351142 Chr.8: 125043949-125043949 on Build GRCh38
rs757021952 Chr.8: 125043914-125043914 on Build GRCh38
rs768045459 Chr.8: 125043860-125043860 on Build GRCh38
rs764828347 Chr.8: 125043899-125043899 on Build GRCh38
rs769830470 Chr.8: 125043834-125043834 on Build GRCh38
rs199604847 Chr.8: 125043943-125043943 on Build GRCh38
rs777035089 Chr.8: 125043842-125043842 on Build GRCh38
rs756271185 Chr.8: 125043844-125043859 on Build GRCh38
rs752193749 Chr.8: 125043902-125043902 on Build GRCh38
rs770743674 Chr.8: 125043821-125043821 on Build GRCh38
rs780129491 Chr.8: 125043857-125043859 on Build GRCh38
rs772106188 Chr.8: 125043817-125043817 on Build GRCh38
rs142685897 Chr.8: 125043911-125043911 on Build GRCh38
rs113111662 Chr.8: 125043843-125043843 on Build GRCh38
rs374415916 Chr.8: 125043957-125043957 on Build GRCh38
rs762452131 Chr.8: 125043966-125043966 on Build GRCh38
rs746082942 Chr.8: 125043933-125043933 on Build GRCh38
rs374702872 Chr.8: 125043824-125043824 on Build GRCh38
rs770055248 Chr.8: 125043942-125043942 on Build GRCh38
rs376039759 Chr.8: 125043904-125043904 on Build GRCh38
rs774756574 Chr.8: 125043849-125043849 on Build GRCh38
rs534139966 Chr.8: 125043839-125043839 on Build GRCh38
rs1019789388 Chr.8: 125043971-125043971 on Build GRCh38
rs769476066 Chr.8: 125043940-125043940 on Build GRCh38
rs781002309 Chr.8: 125043920-125043920 on Build GRCh38
rs545318648 Chr.8: 125043993-125043993 on Build GRCh38
rs757850800 Chr.8: 125043905-125043905 on Build GRCh38
rs1010483156 Chr.8: 125043830-125043830 on Build GRCh38
rs369339497 Chr.8: 125043846-125043846 on Build GRCh38
rs768643408 Chr.8: 125043961-125043961 on Build GRCh38
rs371095857 Chr.8: 125043948-125043948 on Build GRCh38
rs72537275 Chr.8: 125043972-125043973 on Build GRCh38
rs371156369 Chr.8: 125043965-125043965 on Build GRCh38
rs769145000 Chr.8: 125043840-125043840 on Build GRCh38
rs377646434 Chr.8: 125043923-125043923 on Build GRCh38
rs541330058 Chr.8: 125043878-125043878 on Build GRCh38
rs754745588 Chr.8: 125043888-125043888 on Build GRCh38

View Full Product Details