Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 20729967-20730238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP4
Entrez Gene ID
80333
Gene Name
potassium voltage-gated channel interacting protein 4
Gene Aliases
CALP, KCHIP4
UniGene
Hs.655705
Gene Chromosome Location
Chr.4: 20728520-21948801 on Build GRCh38
Transcripts
NM_001035003.1 NM_001035004.1 NM_025221.5 NM_147181.3 NM_147182.3 NM_147183.3 XM_005248190.4 XM_011513882.2 XM_011513883.2 XM_011513884.2 XM_011513885.2 XM_011513886.2 XM_011513887.2 XM_011513889.2 XM_017008653.1 NR_133933.1 XM_011513786.2 XM_011513805.2 XM_017007747.1 XM_017007748.1 XM_017007749.1 XM_017007751.1 XM_017007752.1 XM_017007753.1 XM_017007754.1 XM_017007755.1 XM_017007756.1 XM_017007757.1 XM_017007758.1 XM_017007759.1

Gene Symbol
PACRGL
Entrez Gene ID
133015
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
UniGene
Hs.479298
Gene Chromosome Location
Chr.4: 20696282-20757766 on Build GRCh38
Transcripts
NM_001035003.1 NM_001035004.1 NM_025221.5 NM_147181.3 NM_147182.3 NM_147183.3 XM_005248190.4 XM_011513882.2 XM_011513883.2 XM_011513884.2 XM_011513885.2 XM_011513886.2 XM_011513887.2 XM_011513889.2 XM_017008653.1 NR_133933.1 XM_011513786.2 XM_011513805.2 XM_017007747.1 XM_017007748.1 XM_017007749.1 XM_017007751.1 XM_017007752.1 XM_017007753.1 XM_017007754.1 XM_017007755.1 XM_017007756.1 XM_017007757.1 XM_017007758.1 XM_017007759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3132595 c.604-2A>T p.? Chr.4: 20730131-20730131 on Build GRCh38
1729384 c.724T>A p.S242T Chr.4: 20730111-20730111 on Build GRCh38
1054504 c.619C>T p.R207C Chr.4: 20730114-20730114 on Build GRCh38
3132592 c.706-2A>T p.? Chr.4: 20730131-20730131 on Build GRCh38
1729387 c.622T>A p.S208T Chr.4: 20730111-20730111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029202773 Chr.4: 20730190-20730190 on Build GRCh38
rs201103982 Chr.4: 20730047-20730047 on Build GRCh38
rs564294322 Chr.4: 20730176-20730176 on Build GRCh38
rs954709895 Chr.4: 20730034-20730034 on Build GRCh38
rs765980634 Chr.4: 20730039-20730039 on Build GRCh38
rs377708036 Chr.4: 20730147-20730147 on Build GRCh38
rs12643980 Chr.4: 20730065-20730065 on Build GRCh38
rs377682600 Chr.4: 20730056-20730056 on Build GRCh38
rs762971898 Chr.4: 20730174-20730174 on Build GRCh38
rs149977507 Chr.4: 20730113-20730113 on Build GRCh38
rs1030760409 Chr.4: 20730022-20730022 on Build GRCh38
rs996020954 Chr.4: 20730189-20730189 on Build GRCh38
rs373571715 Chr.4: 20730017-20730017 on Build GRCh38
rs993750985 Chr.4: 20730001-20730001 on Build GRCh38
rs748448529 Chr.4: 20730146-20730146 on Build GRCh38
rs768990870 Chr.4: 20730136-20730136 on Build GRCh38
rs757556154 Chr.4: 20730103-20730103 on Build GRCh38
rs779999949 Chr.4: 20730130-20730131 on Build GRCh38
rs989350897 Chr.4: 20730072-20730072 on Build GRCh38
rs771966590 Chr.4: 20730096-20730097 on Build GRCh38
rs149197141 Chr.4: 20730114-20730114 on Build GRCh38
rs905611878 Chr.4: 20730030-20730030 on Build GRCh38
rs759116590 Chr.4: 20730152-20730152 on Build GRCh38
rs373590424 Chr.4: 20730178-20730178 on Build GRCh38
rs533708266 Chr.4: 20730211-20730211 on Build GRCh38
rs770588421 Chr.4: 20730148-20730148 on Build GRCh38
rs758991826 Chr.4: 20730042-20730042 on Build GRCh38
rs776257365 Chr.4: 20730152-20730153 on Build GRCh38
rs746492253 Chr.4: 20730134-20730135 on Build GRCh38
rs762698250 Chr.4: 20730060-20730060 on Build GRCh38
rs891441557 Chr.4: 20730137-20730137 on Build GRCh38
rs768204165 Chr.4: 20730140-20730141 on Build GRCh38
rs1017665332 Chr.4: 20730059-20730059 on Build GRCh38
rs984141047 Chr.4: 20730192-20730192 on Build GRCh38
rs954136792 Chr.4: 20730191-20730191 on Build GRCh38
rs745686674 Chr.4: 20730037-20730038 on Build GRCh38
rs749777299 Chr.4: 20730134-20730134 on Build GRCh38
rs1008057829 Chr.4: 20730055-20730055 on Build GRCh38
rs769494876 Chr.4: 20730177-20730178 on Build GRCh38
rs78290805 Chr.4: 20730140-20730140 on Build GRCh38
rs769319057 Chr.4: 20730155-20730155 on Build GRCh38
rs777635606 Chr.4: 20730067-20730067 on Build GRCh38
rs186094219 Chr.4: 20730214-20730214 on Build GRCh38
rs200202411 Chr.4: 20730151-20730151 on Build GRCh38
rs747825231 Chr.4: 20730154-20730155 on Build GRCh38
rs781507352 Chr.4: 20730112-20730112 on Build GRCh38
rs763870429 Chr.4: 20730175-20730175 on Build GRCh38
rs771856602 Chr.4: 20730071-20730072 on Build GRCh38
rs774523821 Chr.4: 20730181-20730181 on Build GRCh38
rs967043483 Chr.4: 20730144-20730144 on Build GRCh38
rs901703838 Chr.4: 20730188-20730188 on Build GRCh38
rs370471191 Chr.4: 20730168-20730168 on Build GRCh38

View Full Product Details