Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48688587-48688828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_016291.3 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595119 c.900C>T p.R300R Chr.3: 48688654-48688654 on Build GRCh38
6686595 c.967T>C p.S323P Chr.3: 48688587-48688587 on Build GRCh38
5476463 c.947C>T p.A316V Chr.3: 48688607-48688607 on Build GRCh38
6097821 c.876C>T p.F292F Chr.3: 48688678-48688678 on Build GRCh38
4118328 c.913G>T p.G305C Chr.3: 48688641-48688641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771127659 Chr.3: 48688665-48688665 on Build GRCh38
rs747728098 Chr.3: 48688746-48688746 on Build GRCh38
rs750586432 Chr.3: 48688681-48688681 on Build GRCh38
rs753998544 Chr.3: 48688669-48688669 on Build GRCh38
rs754258572 Chr.3: 48688786-48688786 on Build GRCh38
rs776248772 Chr.3: 48688630-48688630 on Build GRCh38
rs772703699 Chr.3: 48688657-48688657 on Build GRCh38
rs927416196 Chr.3: 48688651-48688651 on Build GRCh38
rs997321233 Chr.3: 48688612-48688612 on Build GRCh38
rs766330061 Chr.3: 48688780-48688783 on Build GRCh38
rs758660945 Chr.3: 48688808-48688808 on Build GRCh38
rs780955935 Chr.3: 48688771-48688771 on Build GRCh38
rs779200655 Chr.3: 48688674-48688674 on Build GRCh38
rs769513321 Chr.3: 48688653-48688653 on Build GRCh38
rs536040651 Chr.3: 48688781-48688781 on Build GRCh38
rs775420494 Chr.3: 48688623-48688623 on Build GRCh38
rs199770977 Chr.3: 48688708-48688708 on Build GRCh38
rs966293134 Chr.3: 48688606-48688606 on Build GRCh38
rs150229636 Chr.3: 48688737-48688737 on Build GRCh38
rs368740167 Chr.3: 48688652-48688652 on Build GRCh38
rs141913549 Chr.3: 48688747-48688747 on Build GRCh38
rs972998281 Chr.3: 48688806-48688806 on Build GRCh38
rs1004832838 Chr.3: 48688790-48688791 on Build GRCh38
rs201907257 Chr.3: 48688784-48688784 on Build GRCh38
rs747008986 Chr.3: 48688625-48688625 on Build GRCh38
rs375983230 Chr.3: 48688619-48688619 on Build GRCh38
rs771788541 Chr.3: 48688620-48688620 on Build GRCh38
rs768689853 Chr.3: 48688728-48688728 on Build GRCh38
rs945679630 Chr.3: 48688726-48688726 on Build GRCh38
rs369147001 Chr.3: 48688803-48688803 on Build GRCh38
rs1043229533 Chr.3: 48688729-48688729 on Build GRCh38
rs376702328 Chr.3: 48688776-48688776 on Build GRCh38
rs770700361 Chr.3: 48688780-48688780 on Build GRCh38
rs138795713 Chr.3: 48688663-48688663 on Build GRCh38
rs372399821 Chr.3: 48688671-48688671 on Build GRCh38
rs759980196 Chr.3: 48688659-48688659 on Build GRCh38
rs775830703 Chr.3: 48688798-48688799 on Build GRCh38
rs750500428 Chr.3: 48688804-48688804 on Build GRCh38
rs768475478 Chr.3: 48688628-48688628 on Build GRCh38
rs761329817 Chr.3: 48688664-48688664 on Build GRCh38
rs746778515 Chr.3: 48688711-48688711 on Build GRCh38
rs761966668 Chr.3: 48688791-48688791 on Build GRCh38
rs758607679 Chr.3: 48688684-48688684 on Build GRCh38

View Full Product Details