Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92240883-92241150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4375759 c.404A>G p.Y135C Chr.9: 92240894-92240894 on Build GRCh38
5968167 c.382C>A p.L128I Chr.9: 92240916-92240916 on Build GRCh38
1210204 c.3183G>T p.K1061N Chr.9: 92240956-92240956 on Build GRCh38
3659501 c.381C>T p.S127S Chr.9: 92240917-92240917 on Build GRCh38
5968166 c.3223C>A p.L1075I Chr.9: 92240916-92240916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747137603 Chr.9: 92241069-92241069 on Build GRCh38
rs550830895 Chr.9: 92241019-92241019 on Build GRCh38
rs568318028 Chr.9: 92240932-92240932 on Build GRCh38
rs766016986 Chr.9: 92240971-92240971 on Build GRCh38
rs776570033 Chr.9: 92240949-92240949 on Build GRCh38
rs371678044 Chr.9: 92240969-92240969 on Build GRCh38
rs754452383 Chr.9: 92240978-92240978 on Build GRCh38
rs749618886 Chr.9: 92241013-92241013 on Build GRCh38
rs750270216 Chr.9: 92240967-92240967 on Build GRCh38
rs895577471 Chr.9: 92241050-92241050 on Build GRCh38
rs112284692 Chr.9: 92240970-92240970 on Build GRCh38
rs764990499 Chr.9: 92240962-92240962 on Build GRCh38
rs771326067 Chr.9: 92240929-92240929 on Build GRCh38
rs745509732 Chr.9: 92240985-92240988 on Build GRCh38
rs772046234 Chr.9: 92240947-92240947 on Build GRCh38
rs1043015907 Chr.9: 92240917-92240917 on Build GRCh38
rs199645059 Chr.9: 92240961-92240961 on Build GRCh38
rs762976612 Chr.9: 92241037-92241037 on Build GRCh38
rs925670441 Chr.9: 92241020-92241020 on Build GRCh38
rs751178239 Chr.9: 92240977-92240977 on Build GRCh38
rs535481901 Chr.9: 92240979-92240979 on Build GRCh38
rs946022681 Chr.9: 92241000-92241000 on Build GRCh38
rs778257886 Chr.9: 92240997-92240997 on Build GRCh38
rs759618440 Chr.9: 92240936-92240936 on Build GRCh38
rs937002019 Chr.9: 92241022-92241022 on Build GRCh38
rs149779047 Chr.9: 92240918-92240918 on Build GRCh38
rs949775286 Chr.9: 92241089-92241089 on Build GRCh38
rs753251179 Chr.9: 92240993-92240993 on Build GRCh38
rs979068255 Chr.9: 92241005-92241005 on Build GRCh38
rs998551447 Chr.9: 92241039-92241039 on Build GRCh38
rs1013071497 Chr.9: 92241096-92241096 on Build GRCh38
rs1057183768 Chr.9: 92241048-92241050 on Build GRCh38
rs1055444484 Chr.9: 92241048-92241048 on Build GRCh38

View Full Product Details