Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208239004-208239277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH1
Entrez Gene ID
3417
Gene Name
isocitrate dehydrogenase (NADP(+)) 1, cytosolic
Gene Aliases
HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD
UniGene
Hs.593422
Gene Chromosome Location
Chr.2: 208236227-208255143 on Build GRCh38
Transcripts
NM_001282386.1 NM_001282387.1 NM_005896.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325527 c.1138A>T p.I380F Chr.2: 208239087-208239087 on Build GRCh38
6979528 c.1046A>G p.N349S Chr.2: 208239179-208239179 on Build GRCh38
6926764 c.1013G>A p.R338K Chr.2: 208239212-208239212 on Build GRCh38
6920421 c.1018T>G p.L340V Chr.2: 208239207-208239207 on Build GRCh38
4774900 c.1110C>T p.G370G Chr.2: 208239115-208239115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201712733 Chr.2: 208239246-208239246 on Build GRCh38
rs763478345 Chr.2: 208239222-208239222 on Build GRCh38
rs765282142 Chr.2: 208239110-208239110 on Build GRCh38
rs201267625 Chr.2: 208239105-208239105 on Build GRCh38
rs775623666 Chr.2: 208239072-208239072 on Build GRCh38
rs745684377 Chr.2: 208239241-208239242 on Build GRCh38
rs752187592 Chr.2: 208239119-208239119 on Build GRCh38
rs777025307 Chr.2: 208239060-208239060 on Build GRCh38
rs760003909 Chr.2: 208239062-208239062 on Build GRCh38
rs200718328 Chr.2: 208239181-208239181 on Build GRCh38
rs186944298 Chr.2: 208239057-208239057 on Build GRCh38
rs758405875 Chr.2: 208239220-208239220 on Build GRCh38
rs371837057 Chr.2: 208239044-208239044 on Build GRCh38
rs746615557 Chr.2: 208239127-208239127 on Build GRCh38
rs775893896 Chr.2: 208239149-208239149 on Build GRCh38
rs756940135 Chr.2: 208239130-208239130 on Build GRCh38
rs766246232 Chr.2: 208239092-208239092 on Build GRCh38
rs762867621 Chr.2: 208239150-208239150 on Build GRCh38
rs775128585 Chr.2: 208239096-208239096 on Build GRCh38
rs764338499 Chr.2: 208239074-208239074 on Build GRCh38
rs770226824 Chr.2: 208239138-208239138 on Build GRCh38
rs752595957 Chr.2: 208239211-208239211 on Build GRCh38
rs776513405 Chr.2: 208239173-208239173 on Build GRCh38
rs751147148 Chr.2: 208239242-208239242 on Build GRCh38
rs777431979 Chr.2: 208239126-208239126 on Build GRCh38
rs139521409 Chr.2: 208239246-208239246 on Build GRCh38
rs766547920 Chr.2: 208239153-208239153 on Build GRCh38
rs200591678 Chr.2: 208239125-208239125 on Build GRCh38
rs762143982 Chr.2: 208239084-208239084 on Build GRCh38
rs369234651 Chr.2: 208239106-208239106 on Build GRCh38
rs200101671 Chr.2: 208239239-208239239 on Build GRCh38
rs536614679 Chr.2: 208239066-208239066 on Build GRCh38
rs372711897 Chr.2: 208239135-208239135 on Build GRCh38
rs199743726 Chr.2: 208239054-208239054 on Build GRCh38
rs762954264 Chr.2: 208239073-208239073 on Build GRCh38
rs141895800 Chr.2: 208239076-208239076 on Build GRCh38
rs536880332 Chr.2: 208239165-208239165 on Build GRCh38
rs112625205 Chr.2: 208239024-208239024 on Build GRCh38
rs771701369 Chr.2: 208239046-208239046 on Build GRCh38
rs754957038 Chr.2: 208239107-208239107 on Build GRCh38
rs763941915 Chr.2: 208239028-208239029 on Build GRCh38
rs375389985 Chr.2: 208239065-208239065 on Build GRCh38
rs759413310 Chr.2: 208239188-208239188 on Build GRCh38
rs750457328 Chr.2: 208239064-208239065 on Build GRCh38
rs751745975 Chr.2: 208239245-208239246 on Build GRCh38
rs768596300 Chr.2: 208239158-208239158 on Build GRCh38
rs781533021 Chr.2: 208239131-208239131 on Build GRCh38
rs765179388 Chr.2: 208239203-208239203 on Build GRCh38
rs763043045 Chr.2: 208239081-208239082 on Build GRCh38
rs367959618 Chr.2: 208239030-208239030 on Build GRCh38
rs376666493 Chr.2: 208239045-208239045 on Build GRCh38

View Full Product Details