Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226795162-226795397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956694 c.3478C>T p.P1160S Chr.2: 226795261-226795261 on Build GRCh38
4952221 c.3354C>A p.P1118P Chr.2: 226795385-226795385 on Build GRCh38
6974851 c.3491C>T p.A1164V Chr.2: 226795248-226795248 on Build GRCh38
6973218 c.3543C>G p.D1181E Chr.2: 226795196-226795196 on Build GRCh38
6090734 c.3540A>G p.I1180M Chr.2: 226795199-226795199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771011699 Chr.2: 226795304-226795304 on Build GRCh38
rs774321419 Chr.2: 226795184-226795184 on Build GRCh38
rs767163776 Chr.2: 226795349-226795349 on Build GRCh38
rs557636520 Chr.2: 226795265-226795265 on Build GRCh38
rs747114219 Chr.2: 226795185-226795185 on Build GRCh38
rs766903970 Chr.2: 226795285-226795285 on Build GRCh38
rs767474165 Chr.2: 226795333-226795334 on Build GRCh38
rs946459210 Chr.2: 226795292-226795292 on Build GRCh38
rs767082046 Chr.2: 226795267-226795267 on Build GRCh38
rs145977916 Chr.2: 226795351-226795351 on Build GRCh38
rs892218709 Chr.2: 226795291-226795291 on Build GRCh38
rs766003841 Chr.2: 226795352-226795352 on Build GRCh38
rs3731594 Chr.2: 226795330-226795330 on Build GRCh38
rs752662327 Chr.2: 226795359-226795359 on Build GRCh38
rs533260174 Chr.2: 226795358-226795358 on Build GRCh38
rs775770356 Chr.2: 226795320-226795320 on Build GRCh38
rs899984257 Chr.2: 226795327-226795327 on Build GRCh38
rs947989412 Chr.2: 226795231-226795231 on Build GRCh38
rs759707132 Chr.2: 226795220-226795220 on Build GRCh38
rs765622405 Chr.2: 226795234-226795234 on Build GRCh38
rs780147733 Chr.2: 226795376-226795376 on Build GRCh38
rs753853576 Chr.2: 226795334-226795336 on Build GRCh38
rs148833195 Chr.2: 226795288-226795288 on Build GRCh38
rs112888661 Chr.2: 226795374-226795374 on Build GRCh38
rs776841403 Chr.2: 226795206-226795206 on Build GRCh38
rs763295813 Chr.2: 226795321-226795321 on Build GRCh38
rs745918297 Chr.2: 226795316-226795316 on Build GRCh38
rs202089976 Chr.2: 226795258-226795258 on Build GRCh38
rs770993310 Chr.2: 226795200-226795200 on Build GRCh38
rs758373069 Chr.2: 226795367-226795367 on Build GRCh38
rs143913301 Chr.2: 226795377-226795377 on Build GRCh38
rs759343037 Chr.2: 226795225-226795233 on Build GRCh38
rs200821911 Chr.2: 226795309-226795309 on Build GRCh38
rs764570043 Chr.2: 226795257-226795257 on Build GRCh38
rs553359474 Chr.2: 226795224-226795224 on Build GRCh38
rs573529594 Chr.2: 226795331-226795331 on Build GRCh38
rs1043936986 Chr.2: 226795323-226795323 on Build GRCh38
rs754786896 Chr.2: 226795287-226795287 on Build GRCh38
rs576198727 Chr.2: 226795272-226795272 on Build GRCh38
rs200142054 Chr.2: 226795333-226795333 on Build GRCh38
rs748011125 Chr.2: 226795299-226795299 on Build GRCh38
rs749083567 Chr.2: 226795280-226795280 on Build GRCh38
rs372697297 Chr.2: 226795269-226795269 on Build GRCh38
rs777666532 Chr.2: 226795372-226795372 on Build GRCh38
rs765435936 Chr.2: 226795374-226795379 on Build GRCh38
rs143317854 Chr.2: 226795250-226795250 on Build GRCh38
rs770051216 Chr.2: 226795317-226795317 on Build GRCh38
rs763355412 Chr.2: 226795255-226795255 on Build GRCh38
rs371306259 Chr.2: 226795357-226795357 on Build GRCh38
rs750119405 Chr.2: 226795268-226795268 on Build GRCh38

View Full Product Details