Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184418525-184418774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2
Entrez Gene ID
3660
Gene Name
interferon regulatory factor 2
Gene Aliases
IRF-2
UniGene
Hs.654566
Gene Chromosome Location
Chr.4: 184387722-184474572 on Build GRCh38
Transcripts
NM_002199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5803739 c.364+1G>A p.? Chr.4: 184418531-184418531 on Build GRCh38
5866126 c.352C>T p.P118S Chr.4: 184418544-184418544 on Build GRCh38
6166916 c.238A>T p.N80Y Chr.4: 184418658-184418658 on Build GRCh38
3696566 c.339C>T p.P113P Chr.4: 184418557-184418557 on Build GRCh38
6586312 c.198delA p.Q66fs*4 Chr.4: 184418698-184418698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757515001 Chr.4: 184418743-184418743 on Build GRCh38
rs760988459 Chr.4: 184418697-184418697 on Build GRCh38
rs370144090 Chr.4: 184418650-184418650 on Build GRCh38
rs994374555 Chr.4: 184418738-184418738 on Build GRCh38
rs777421604 Chr.4: 184418737-184418738 on Build GRCh38
rs111680592 Chr.4: 184418737-184418737 on Build GRCh38
rs776421591 Chr.4: 184418742-184418742 on Build GRCh38
rs148758064 Chr.4: 184418647-184418647 on Build GRCh38
rs144286568 Chr.4: 184418557-184418557 on Build GRCh38
rs768241157 Chr.4: 184418584-184418584 on Build GRCh38
rs927732513 Chr.4: 184418554-184418554 on Build GRCh38
rs200277901 Chr.4: 184418674-184418674 on Build GRCh38
rs548075592 Chr.4: 184418742-184418742 on Build GRCh38
rs769820948 Chr.4: 184418721-184418721 on Build GRCh38
rs745405030 Chr.4: 184418550-184418550 on Build GRCh38
rs1035992628 Chr.4: 184418715-184418715 on Build GRCh38
rs760665800 Chr.4: 184418619-184418619 on Build GRCh38
rs747859951 Chr.4: 184418629-184418629 on Build GRCh38
rs746653734 Chr.4: 184418559-184418559 on Build GRCh38
rs199802618 Chr.4: 184418718-184418718 on Build GRCh38
rs776493551 Chr.4: 184418623-184418623 on Build GRCh38
rs762752435 Chr.4: 184418659-184418659 on Build GRCh38
rs4861623 Chr.4: 184418740-184418740 on Build GRCh38
rs941241429 Chr.4: 184418729-184418729 on Build GRCh38
rs767697198 Chr.4: 184418694-184418694 on Build GRCh38
rs775910534 Chr.4: 184418696-184418696 on Build GRCh38
rs146592173 Chr.4: 184418638-184418638 on Build GRCh38

View Full Product Details