Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 178828133-178828406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB2
Entrez Gene ID
10242
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 2
Gene Aliases
-
UniGene
Hs.478368
Gene Chromosome Location
Chr.3: 178536298-178844429 on Build GRCh38
Transcripts
NM_001278911.1 NM_005832.4 NM_181361.2 XM_011512325.2 NR_126560.1 NR_126561.1

Gene Symbol
KCNMB2-AS1
Entrez Gene ID
104797538
Gene Name
KCNMB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.478363
Gene Chromosome Location
Chr.3: 178525467-178860405 on Build GRCh38
Transcripts
NM_001278911.1 NM_005832.4 NM_181361.2 XM_011512325.2 NR_126560.1 NR_126561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4763688 c.356A>G p.N119S Chr.3: 178828306-178828306 on Build GRCh38
3590397 c.293C>T p.S98F Chr.3: 178828243-178828243 on Build GRCh38
7329746 c.391C>A p.H131N Chr.3: 178828341-178828341 on Build GRCh38
1041428 c.238G>A p.E80K Chr.3: 178828188-178828188 on Build GRCh38
3590398 c.342C>T p.L114L Chr.3: 178828292-178828292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763453145 Chr.3: 178828329-178828329 on Build GRCh38
rs1043669736 Chr.3: 178828369-178828369 on Build GRCh38
rs536282066 Chr.3: 178828161-178828161 on Build GRCh38
rs765629089 Chr.3: 178828222-178828222 on Build GRCh38
rs764639111 Chr.3: 178828214-178828214 on Build GRCh38
rs373338689 Chr.3: 178828319-178828319 on Build GRCh38
rs145640012 Chr.3: 178828192-178828192 on Build GRCh38
rs757687037 Chr.3: 178828220-178828220 on Build GRCh38
rs745693456 Chr.3: 178828295-178828295 on Build GRCh38
rs777753416 Chr.3: 178828251-178828251 on Build GRCh38
rs573122943 Chr.3: 178828216-178828216 on Build GRCh38
rs201338193 Chr.3: 178828320-178828320 on Build GRCh38
rs749289810 Chr.3: 178828252-178828252 on Build GRCh38
rs750779813 Chr.3: 178828323-178828323 on Build GRCh38
rs946980945 Chr.3: 178828328-178828328 on Build GRCh38
rs35616186 Chr.3: 178828164-178828164 on Build GRCh38
rs749090848 Chr.3: 178828168-178828168 on Build GRCh38
rs563719552 Chr.3: 178828188-178828188 on Build GRCh38
rs754356731 Chr.3: 178828217-178828217 on Build GRCh38
rs368918405 Chr.3: 178828228-178828228 on Build GRCh38
rs773067094 Chr.3: 178828165-178828165 on Build GRCh38
rs905232062 Chr.3: 178828381-178828381 on Build GRCh38
rs750828534 Chr.3: 178828225-178828225 on Build GRCh38
rs575957428 Chr.3: 178828322-178828322 on Build GRCh38
rs376796126 Chr.3: 178828179-178828179 on Build GRCh38
rs868351449 Chr.3: 178828276-178828276 on Build GRCh38
rs781066708 Chr.3: 178828380-178828380 on Build GRCh38
rs751790073 Chr.3: 178828355-178828355 on Build GRCh38
rs9831934 Chr.3: 178828238-178828238 on Build GRCh38
rs760218747 Chr.3: 178828307-178828307 on Build GRCh38
rs770862207 Chr.3: 178828271-178828271 on Build GRCh38
rs752789139 Chr.3: 178828385-178828385 on Build GRCh38
rs771678569 Chr.3: 178828301-178828301 on Build GRCh38
rs747083799 Chr.3: 178828163-178828163 on Build GRCh38
rs758647206 Chr.3: 178828226-178828226 on Build GRCh38
rs766606256 Chr.3: 178828345-178828345 on Build GRCh38
rs767171556 Chr.3: 178828187-178828187 on Build GRCh38
rs755065068 Chr.3: 178828241-178828241 on Build GRCh38
rs575178662 Chr.3: 178828302-178828302 on Build GRCh38
rs770772310 Chr.3: 178828178-178828178 on Build GRCh38
rs774075687 Chr.3: 178828294-178828294 on Build GRCh38
rs755082250 Chr.3: 178828377-178828377 on Build GRCh38
rs751001749 Chr.3: 178828273-178828274 on Build GRCh38

View Full Product Details