Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18436159-18436394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4693195 c.812A>C p.E271A Chr.19: 18436195-18436195 on Build GRCh38
6687659 c.736G>A p.E246K Chr.19: 18436353-18436353 on Build GRCh38
3100062 c.793G>A p.A265T Chr.19: 18436214-18436214 on Build GRCh38
6585866 c.734C>G p.A245G Chr.19: 18436355-18436355 on Build GRCh38
7272276 c.759+10C>T p.? Chr.19: 18436320-18436320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200994797 Chr.19: 18436221-18436221 on Build GRCh38
rs187427629 Chr.19: 18436255-18436255 on Build GRCh38
rs769489890 Chr.19: 18436227-18436227 on Build GRCh38
rs865897417 Chr.19: 18436285-18436286 on Build GRCh38
rs762119762 Chr.19: 18436289-18436290 on Build GRCh38
rs745562139 Chr.19: 18436306-18436306 on Build GRCh38
rs140705791 Chr.19: 18436177-18436177 on Build GRCh38
rs546911488 Chr.19: 18436254-18436254 on Build GRCh38
rs780982383 Chr.19: 18436300-18436300 on Build GRCh38
rs776787019 Chr.19: 18436368-18436368 on Build GRCh38
rs919183477 Chr.19: 18436258-18436258 on Build GRCh38
rs1056795336 Chr.19: 18436248-18436248 on Build GRCh38
rs777377120 Chr.19: 18436272-18436272 on Build GRCh38
rs138624155 Chr.19: 18436373-18436373 on Build GRCh38
rs112525554 Chr.19: 18436287-18436288 on Build GRCh38
rs769124619 Chr.19: 18436281-18436284 on Build GRCh38
rs371984676 Chr.19: 18436351-18436351 on Build GRCh38
rs765710672 Chr.19: 18436194-18436194 on Build GRCh38
rs750708791 Chr.19: 18436253-18436253 on Build GRCh38
rs766323136 Chr.19: 18436363-18436363 on Build GRCh38
rs200357380 Chr.19: 18436341-18436341 on Build GRCh38
rs762735088 Chr.19: 18436371-18436371 on Build GRCh38
rs753244762 Chr.19: 18436230-18436230 on Build GRCh38
rs769393805 Chr.19: 18436307-18436307 on Build GRCh38
rs769218905 Chr.19: 18436327-18436327 on Build GRCh38
rs752538721 Chr.19: 18436257-18436257 on Build GRCh38
rs200198473 Chr.19: 18436315-18436315 on Build GRCh38
rs369772066 Chr.19: 18436277-18436277 on Build GRCh38
rs766522486 Chr.19: 18436256-18436256 on Build GRCh38
rs984105080 Chr.19: 18436279-18436279 on Build GRCh38
rs758383418 Chr.19: 18436260-18436260 on Build GRCh38
rs986311464 Chr.19: 18436205-18436205 on Build GRCh38
rs773393316 Chr.19: 18436345-18436345 on Build GRCh38
rs757190038 Chr.19: 18436289-18436289 on Build GRCh38
rs375448110 Chr.19: 18436360-18436360 on Build GRCh38
rs774511397 Chr.19: 18436333-18436333 on Build GRCh38
rs746594892 Chr.19: 18436193-18436193 on Build GRCh38
rs150177719 Chr.19: 18436215-18436215 on Build GRCh38
rs896440976 Chr.19: 18436249-18436249 on Build GRCh38
rs564464594 Chr.19: 18436280-18436280 on Build GRCh38
rs371293031 Chr.19: 18436239-18436239 on Build GRCh38
rs375525635 Chr.19: 18436243-18436243 on Build GRCh38
rs749706847 Chr.19: 18436319-18436319 on Build GRCh38
rs544339704 Chr.19: 18436185-18436185 on Build GRCh38
rs1056318709 Chr.19: 18436342-18436342 on Build GRCh38
rs762160711 Chr.19: 18436334-18436334 on Build GRCh38
rs761965694 Chr.19: 18436241-18436241 on Build GRCh38
rs768859602 Chr.19: 18436231-18436231 on Build GRCh38
rs777655216 Chr.19: 18436191-18436191 on Build GRCh38

View Full Product Details