Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43292719-43292976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCS1
Entrez Gene ID
3157
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 1
Gene Aliases
HMGCS
UniGene
Hs.397729
Gene Chromosome Location
Chr.5: 43287470-43313512 on Build GRCh38
Transcripts
NM_001098272.2 NM_001324219.1 NM_001324220.1 NM_001324222.1 NM_001324223.1 NM_001324224.1 NM_002130.7 XM_011514036.2 XM_017009412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241999 c.1291G>A p.E431K Chr.5: 43292866-43292866 on Build GRCh38
6513042 c.1210A>T p.S404C Chr.5: 43292947-43292947 on Build GRCh38
1068403 c.1270G>A p.A424T Chr.5: 43292887-43292887 on Build GRCh38
4523524 c.1219G>A p.D407N Chr.5: 43292938-43292938 on Build GRCh38
1068404 c.1269C>T p.F423F Chr.5: 43292888-43292888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930285262 Chr.5: 43292817-43292817 on Build GRCh38
rs1018652181 Chr.5: 43292904-43292904 on Build GRCh38
rs749825410 Chr.5: 43292836-43292836 on Build GRCh38
rs892276353 Chr.5: 43292909-43292909 on Build GRCh38
rs745431136 Chr.5: 43292838-43292838 on Build GRCh38
rs138346407 Chr.5: 43292762-43292762 on Build GRCh38
rs888852464 Chr.5: 43292887-43292887 on Build GRCh38
rs530208538 Chr.5: 43292948-43292948 on Build GRCh38
rs182142394 Chr.5: 43292810-43292810 on Build GRCh38
rs748480296 Chr.5: 43292826-43292826 on Build GRCh38
rs548277878 Chr.5: 43292930-43292930 on Build GRCh38
rs774163663 Chr.5: 43292917-43292917 on Build GRCh38
rs150826961 Chr.5: 43292888-43292888 on Build GRCh38
rs1054443881 Chr.5: 43292778-43292778 on Build GRCh38
rs769345913 Chr.5: 43292862-43292862 on Build GRCh38
rs373702814 Chr.5: 43292825-43292825 on Build GRCh38
rs767403990 Chr.5: 43292936-43292936 on Build GRCh38
rs779440067 Chr.5: 43292818-43292818 on Build GRCh38
rs1009367549 Chr.5: 43292940-43292940 on Build GRCh38
rs756205814 Chr.5: 43292797-43292797 on Build GRCh38
rs112595821 Chr.5: 43292811-43292811 on Build GRCh38
rs772492906 Chr.5: 43292833-43292833 on Build GRCh38
rs763689160 Chr.5: 43292912-43292912 on Build GRCh38
rs147358497 Chr.5: 43292771-43292771 on Build GRCh38
rs773742561 Chr.5: 43292834-43292834 on Build GRCh38
rs879833539 Chr.5: 43292799-43292799 on Build GRCh38
rs149631630 Chr.5: 43292769-43292769 on Build GRCh38
rs55794239 Chr.5: 43292806-43292806 on Build GRCh38
rs750369430 Chr.5: 43292941-43292941 on Build GRCh38
rs762609462 Chr.5: 43292908-43292908 on Build GRCh38
rs1050136237 Chr.5: 43292873-43292873 on Build GRCh38
rs907448761 Chr.5: 43292915-43292915 on Build GRCh38

View Full Product Details