Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135892288-135892561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL9
Entrez Gene ID
3578
Gene Name
interleukin 9
Gene Aliases
HP40, IL-9, P40
UniGene
Hs.960
Gene Chromosome Location
Chr.5: 135892246-135895827 on Build GRCh38
Transcripts
NM_000590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692373 c.361G>A p.A121T Chr.5: 135892465-135892465 on Build GRCh38
1061010 c.392T>C p.I131T Chr.5: 135892434-135892434 on Build GRCh38
6685459 c.363G>A p.A121A Chr.5: 135892463-135892463 on Build GRCh38
3993979 c.391A>G p.I131V Chr.5: 135892435-135892435 on Build GRCh38
6240762 c.351G>C p.T117T Chr.5: 135892475-135892475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766994085 Chr.5: 135892394-135892394 on Build GRCh38
rs573907716 Chr.5: 135892475-135892475 on Build GRCh38
rs770053235 Chr.5: 135892352-135892352 on Build GRCh38
rs201509284 Chr.5: 135892464-135892464 on Build GRCh38
rs777896084 Chr.5: 135892446-135892446 on Build GRCh38
rs769440030 Chr.5: 135892359-135892359 on Build GRCh38
rs764277805 Chr.5: 135892387-135892387 on Build GRCh38
rs967577228 Chr.5: 135892492-135892492 on Build GRCh38
rs768274071 Chr.5: 135892467-135892467 on Build GRCh38
rs758437814 Chr.5: 135892341-135892341 on Build GRCh38
rs140115692 Chr.5: 135892449-135892449 on Build GRCh38
rs141219153 Chr.5: 135892463-135892463 on Build GRCh38
rs761327797 Chr.5: 135892389-135892389 on Build GRCh38
rs752902305 Chr.5: 135892432-135892432 on Build GRCh38
rs751590770 Chr.5: 135892519-135892519 on Build GRCh38
rs775039070 Chr.5: 135892365-135892365 on Build GRCh38
rs775913762 Chr.5: 135892363-135892366 on Build GRCh38
rs765568745 Chr.5: 135892493-135892493 on Build GRCh38
rs2069885 Chr.5: 135892476-135892476 on Build GRCh38
rs768335425 Chr.5: 135892386-135892386 on Build GRCh38
rs763094801 Chr.5: 135892509-135892509 on Build GRCh38
rs764314957 Chr.5: 135892518-135892518 on Build GRCh38
rs761419979 Chr.5: 135892473-135892473 on Build GRCh38
rs777656245 Chr.5: 135892347-135892347 on Build GRCh38
rs371222135 Chr.5: 135892466-135892466 on Build GRCh38
rs202065589 Chr.5: 135892445-135892445 on Build GRCh38
rs765457187 Chr.5: 135892424-135892424 on Build GRCh38
rs762497183 Chr.5: 135892366-135892366 on Build GRCh38
rs201186684 Chr.5: 135892412-135892412 on Build GRCh38
rs752950009 Chr.5: 135892500-135892500 on Build GRCh38
rs759725844 Chr.5: 135892343-135892344 on Build GRCh38
rs781015452 Chr.5: 135892458-135892458 on Build GRCh38
rs534018695 Chr.5: 135892465-135892465 on Build GRCh38
rs770709220 Chr.5: 135892352-135892352 on Build GRCh38
rs757360736 Chr.5: 135892521-135892521 on Build GRCh38
rs200128004 Chr.5: 135892472-135892472 on Build GRCh38
rs181324057 Chr.5: 135892372-135892372 on Build GRCh38
rs759912180 Chr.5: 135892488-135892488 on Build GRCh38
rs776791609 Chr.5: 135892341-135892344 on Build GRCh38
rs754170371 Chr.5: 135892396-135892396 on Build GRCh38
rs527870399 Chr.5: 135892357-135892357 on Build GRCh38
rs751643624 Chr.5: 135892448-135892448 on Build GRCh38
rs746996861 Chr.5: 135892351-135892351 on Build GRCh38
rs970229668 Chr.5: 135892457-135892457 on Build GRCh38
rs780898220 Chr.5: 135892355-135892355 on Build GRCh38

View Full Product Details