Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49153261-49153494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRC
Entrez Gene ID
3270
Gene Name
histidine rich calcium binding protein
Gene Aliases
-
UniGene
Hs.436885
Gene Chromosome Location
Chr.19: 49151198-49155443 on Build GRCh38
Transcripts
NM_002152.2 XM_017026733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630507 c.1889G>A p.C630Y Chr.19: 49153274-49153274 on Build GRCh38
2752185 c.1818C>T p.S606S Chr.19: 49153420-49153420 on Build GRCh38
6892069 c.1786G>A p.E596K Chr.19: 49153452-49153452 on Build GRCh38
6682086 c.1870G>T p.G624W Chr.19: 49153293-49153293 on Build GRCh38
3960270 c.1831+8G>T p.? Chr.19: 49153399-49153399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754673679 Chr.19: 49153375-49153375 on Build GRCh38
rs749435193 Chr.19: 49153389-49153389 on Build GRCh38
rs751388855 Chr.19: 49153437-49153437 on Build GRCh38
rs758380352 Chr.19: 49153371-49153371 on Build GRCh38
rs768861872 Chr.19: 49153419-49153419 on Build GRCh38
rs1006831164 Chr.19: 49153316-49153316 on Build GRCh38
rs778928795 Chr.19: 49153394-49153394 on Build GRCh38
rs770934147 Chr.19: 49153307-49153307 on Build GRCh38
rs140510206 Chr.19: 49153426-49153426 on Build GRCh38
rs999520609 Chr.19: 49153341-49153341 on Build GRCh38
rs201472861 Chr.19: 49153402-49153402 on Build GRCh38
rs375450653 Chr.19: 49153450-49153450 on Build GRCh38
rs781050539 Chr.19: 49153384-49153384 on Build GRCh38
rs761002041 Chr.19: 49153321-49153321 on Build GRCh38
rs533706046 Chr.19: 49153287-49153287 on Build GRCh38
rs772604975 Chr.19: 49153319-49153319 on Build GRCh38
rs988103853 Chr.19: 49153396-49153396 on Build GRCh38
rs375861220 Chr.19: 49153372-49153372 on Build GRCh38
rs373250549 Chr.19: 49153370-49153370 on Build GRCh38
rs775825134 Chr.19: 49153320-49153320 on Build GRCh38
rs766002982 Chr.19: 49153434-49153434 on Build GRCh38
rs745885568 Chr.19: 49153335-49153336 on Build GRCh38
rs539739005 Chr.19: 49153474-49153474 on Build GRCh38
rs764530691 Chr.19: 49153328-49153328 on Build GRCh38
rs952174786 Chr.19: 49153295-49153295 on Build GRCh38
rs373869833 Chr.19: 49153289-49153289 on Build GRCh38
rs761535993 Chr.19: 49153421-49153421 on Build GRCh38
rs757463089 Chr.19: 49153468-49153468 on Build GRCh38
rs774433232 Chr.19: 49153312-49153312 on Build GRCh38
rs914345713 Chr.19: 49153431-49153431 on Build GRCh38
rs113401930 Chr.19: 49153432-49153432 on Build GRCh38
rs759552944 Chr.19: 49153442-49153442 on Build GRCh38
rs751359540 Chr.19: 49153374-49153374 on Build GRCh38
rs775801597 Chr.19: 49153407-49153407 on Build GRCh38
rs190856179 Chr.19: 49153420-49153420 on Build GRCh38
rs757516023 Chr.19: 49153392-49153392 on Build GRCh38
rs76146490 Chr.19: 49153335-49153335 on Build GRCh38
rs745427517 Chr.19: 49153315-49153315 on Build GRCh38
rs61732829 Chr.19: 49153431-49153431 on Build GRCh38
rs781266560 Chr.19: 49153284-49153284 on Build GRCh38
rs371182252 Chr.19: 49153350-49153350 on Build GRCh38
rs200937338 Chr.19: 49153336-49153336 on Build GRCh38
rs1032792023 Chr.19: 49153354-49153354 on Build GRCh38
rs377024240 Chr.19: 49153411-49153411 on Build GRCh38
rs150390369 Chr.19: 49153459-49153459 on Build GRCh38
rs746011475 Chr.19: 49153401-49153401 on Build GRCh38

View Full Product Details