Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132117049-132117284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6889206 c.264G>A p.Q88Q Chr.10: 132117205-132117205 on Build GRCh38
3435761 c.154G>A p.E52K Chr.10: 132117095-132117095 on Build GRCh38
6889205 c.265G>A p.A89T Chr.10: 132117206-132117206 on Build GRCh38
3435762 c.246G>A p.E82E Chr.10: 132117187-132117187 on Build GRCh38
683971 c.271C>T p.R91C Chr.10: 132117212-132117212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371348426 Chr.10: 132117093-132117093 on Build GRCh38
rs1035341272 Chr.10: 132117200-132117200 on Build GRCh38
rs778187502 Chr.10: 132117252-132117252 on Build GRCh38
rs756084381 Chr.10: 132117120-132117120 on Build GRCh38
rs574033707 Chr.10: 132117182-132117182 on Build GRCh38
rs752511214 Chr.10: 132117092-132117092 on Build GRCh38
rs867161908 Chr.10: 132117114-132117114 on Build GRCh38
rs377713578 Chr.10: 132117075-132117075 on Build GRCh38
rs557088199 Chr.10: 132117145-132117145 on Build GRCh38
rs1002560053 Chr.10: 132117178-132117178 on Build GRCh38
rs750533811 Chr.10: 132117113-132117113 on Build GRCh38
rs751521815 Chr.10: 132117191-132117191 on Build GRCh38
rs370393227 Chr.10: 132117147-132117147 on Build GRCh38
rs779055507 Chr.10: 132117220-132117220 on Build GRCh38
rs775450865 Chr.10: 132117171-132117171 on Build GRCh38
rs990367724 Chr.10: 132117240-132117240 on Build GRCh38
rs1016135179 Chr.10: 132117226-132117226 on Build GRCh38
rs753880520 Chr.10: 132117213-132117213 on Build GRCh38
rs374779116 Chr.10: 132117212-132117212 on Build GRCh38
rs201597769 Chr.10: 132117136-132117136 on Build GRCh38
rs199768813 Chr.10: 132117261-132117261 on Build GRCh38
rs375659509 Chr.10: 132117121-132117121 on Build GRCh38
rs377405834 Chr.10: 132117137-132117137 on Build GRCh38
rs367618615 Chr.10: 132117219-132117219 on Build GRCh38
rs759304993 Chr.10: 132117148-132117148 on Build GRCh38
rs199726287 Chr.10: 132117123-132117123 on Build GRCh38
rs762782306 Chr.10: 132117181-132117181 on Build GRCh38
rs963248529 Chr.10: 132117244-132117244 on Build GRCh38
rs777525762 Chr.10: 132117094-132117094 on Build GRCh38
rs757200962 Chr.10: 132117199-132117199 on Build GRCh38
rs779286840 Chr.10: 132117124-132117124 on Build GRCh38
rs770407014 Chr.10: 132117146-132117146 on Build GRCh38
rs1013416975 Chr.10: 132117125-132117125 on Build GRCh38
rs764944954 Chr.10: 132117160-132117160 on Build GRCh38
rs867229121 Chr.10: 132117255-132117255 on Build GRCh38
rs780173989 Chr.10: 132117122-132117122 on Build GRCh38
rs758623280 Chr.10: 132117227-132117227 on Build GRCh38
rs748470939 Chr.10: 132117132-132117132 on Build GRCh38
rs370253977 Chr.10: 132117224-132117224 on Build GRCh38

View Full Product Details