Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128988853-128989086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_020741.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006944.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006948.1 XM_017006949.1 XM_017006950.1 XM_017006951.1 XM_017006952.1 XM_017006953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3587166 c.272G>A p.G91E Chr.3: 128988909-128988909 on Build GRCh38
4539694 c.271G>A p.G91R Chr.3: 128988910-128988910 on Build GRCh38
282171 c.293A>G p.E98G Chr.3: 128988888-128988888 on Build GRCh38
1484579 c.301A>G p.K101E Chr.3: 128988880-128988880 on Build GRCh38
4624952 c.266A>C p.Q89P Chr.3: 128988915-128988915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967757615 Chr.3: 128988969-128988969 on Build GRCh38
rs916409861 Chr.3: 128989022-128989023 on Build GRCh38
rs753264473 Chr.3: 128988896-128988896 on Build GRCh38
rs867991320 Chr.3: 128988902-128988902 on Build GRCh38
rs369202308 Chr.3: 128988897-128988897 on Build GRCh38
rs947848519 Chr.3: 128989036-128989036 on Build GRCh38
rs184740938 Chr.3: 128988949-128988949 on Build GRCh38
rs939088054 Chr.3: 128988980-128988980 on Build GRCh38
rs977699515 Chr.3: 128988971-128988971 on Build GRCh38
rs894276312 Chr.3: 128988918-128988918 on Build GRCh38
rs373049779 Chr.3: 128988909-128988909 on Build GRCh38
rs923722415 Chr.3: 128988974-128988974 on Build GRCh38
rs1011829708 Chr.3: 128988923-128988923 on Build GRCh38
rs548430737 Chr.3: 128988912-128988912 on Build GRCh38
rs765844557 Chr.3: 128988950-128988951 on Build GRCh38
rs376851400 Chr.3: 128988946-128988946 on Build GRCh38
rs757533284 Chr.3: 128988934-128988934 on Build GRCh38
rs752011664 Chr.3: 128988929-128988929 on Build GRCh38
rs749411103 Chr.3: 128988982-128988982 on Build GRCh38
rs998758574 Chr.3: 128989008-128989008 on Build GRCh38
rs781622733 Chr.3: 128988945-128988945 on Build GRCh38

View Full Product Details