Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1519828-1520075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT140
Entrez Gene ID
9742
Gene Name
intraflagellar transport 140
Gene Aliases
MZSDS, SRTD9, WDTC2, c305C8.4, c380F5.1, gs114
UniGene
Hs.389438
Gene Chromosome Location
Chr.16: 1510427-1612108 on Build GRCh38
Transcripts
NM_014714.3 XM_006720990.3 XM_006720991.3 XM_006720992.3 XM_011522766.2 XM_011522767.2 XM_017023910.1 XM_017023911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2126838 c.3980C>T p.T1327I Chr.16: 1519941-1519941 on Build GRCh38
5850230 c.3901G>A p.D1301N Chr.16: 1520020-1520020 on Build GRCh38
1210306 c.3970G>A p.D1324N Chr.16: 1519951-1519951 on Build GRCh38
5581347 c.4026C>T p.F1342F Chr.16: 1519895-1519895 on Build GRCh38
5473690 c.3900C>T p.Y1300Y Chr.16: 1520021-1520021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756142477 Chr.16: 1519996-1519996 on Build GRCh38
rs200699325 Chr.16: 1519933-1519933 on Build GRCh38
rs770059068 Chr.16: 1519977-1519977 on Build GRCh38
rs556735183 Chr.16: 1519885-1519885 on Build GRCh38
rs1055889482 Chr.16: 1519993-1519993 on Build GRCh38
rs765346575 Chr.16: 1519903-1519903 on Build GRCh38
rs148387007 Chr.16: 1520021-1520021 on Build GRCh38
rs147025984 Chr.16: 1520044-1520044 on Build GRCh38
rs753810506 Chr.16: 1519986-1519986 on Build GRCh38
rs373217447 Chr.16: 1520051-1520051 on Build GRCh38
rs562033874 Chr.16: 1520030-1520030 on Build GRCh38
rs765101783 Chr.16: 1519848-1519848 on Build GRCh38
rs746697405 Chr.16: 1519961-1519966 on Build GRCh38
rs759849021 Chr.16: 1519901-1519901 on Build GRCh38
rs751530824 Chr.16: 1519922-1519922 on Build GRCh38
rs745931197 Chr.16: 1519975-1519975 on Build GRCh38
rs775694591 Chr.16: 1519978-1519978 on Build GRCh38
rs761018449 Chr.16: 1519892-1519892 on Build GRCh38
rs774567356 Chr.16: 1519852-1519863 on Build GRCh38
rs749563050 Chr.16: 1520048-1520048 on Build GRCh38
rs907068323 Chr.16: 1519972-1519972 on Build GRCh38
rs755596826 Chr.16: 1520008-1520008 on Build GRCh38
rs942381725 Chr.16: 1519886-1519886 on Build GRCh38
rs763810221 Chr.16: 1519914-1519914 on Build GRCh38
rs776522454 Chr.16: 1519895-1519895 on Build GRCh38
rs747304190 Chr.16: 1520029-1520029 on Build GRCh38
rs538447838 Chr.16: 1519875-1519875 on Build GRCh38
rs754625031 Chr.16: 1520020-1520020 on Build GRCh38
rs794727473 Chr.16: 1519930-1519930 on Build GRCh38
rs371103494 Chr.16: 1519958-1519958 on Build GRCh38
rs750011683 Chr.16: 1520004-1520004 on Build GRCh38
rs138311751 Chr.16: 1520046-1520046 on Build GRCh38
rs144624901 Chr.16: 1519870-1519870 on Build GRCh38
rs539917190 Chr.16: 1520040-1520040 on Build GRCh38
rs61745103 Chr.16: 1519931-1519931 on Build GRCh38
rs377688969 Chr.16: 1519935-1519935 on Build GRCh38
rs1035130199 Chr.16: 1520014-1520014 on Build GRCh38
rs1054102092 Chr.16: 1519899-1519899 on Build GRCh38
rs748523056 Chr.16: 1519880-1519880 on Build GRCh38
rs181794285 Chr.16: 1519955-1519955 on Build GRCh38
rs373703197 Chr.16: 1519866-1519866 on Build GRCh38
rs144050063 Chr.16: 1519884-1519884 on Build GRCh38
rs761853895 Chr.16: 1520001-1520001 on Build GRCh38
rs752553987 Chr.16: 1519906-1519906 on Build GRCh38
rs986239975 Chr.16: 1520045-1520045 on Build GRCh38
rs34985348 Chr.16: 1519989-1519989 on Build GRCh38
rs1002670900 Chr.16: 1519982-1519982 on Build GRCh38
rs574131281 Chr.16: 1519979-1519979 on Build GRCh38
rs762750524 Chr.16: 1519912-1519912 on Build GRCh38
rs376151586 Chr.16: 1519932-1519932 on Build GRCh38
rs374747598 Chr.16: 1519862-1519862 on Build GRCh38
rs138364426 Chr.16: 1519871-1519871 on Build GRCh38
rs779462689 Chr.16: 1519878-1519878 on Build GRCh38
rs767354864 Chr.16: 1520003-1520003 on Build GRCh38
rs772351317 Chr.16: 1519960-1519961 on Build GRCh38
rs587776909 Chr.16: 1520005-1520006 on Build GRCh38
rs758371286 Chr.16: 1519861-1519861 on Build GRCh38
rs375363045 Chr.16: 1520050-1520050 on Build GRCh38
rs1030507437 Chr.16: 1520043-1520043 on Build GRCh38
rs773997219 Chr.16: 1519997-1519997 on Build GRCh38
rs758053488 Chr.16: 1519943-1519943 on Build GRCh38
rs140443906 Chr.16: 1520036-1520036 on Build GRCh38
rs771070134 Chr.16: 1519894-1519894 on Build GRCh38
rs771318853 Chr.16: 1519959-1519959 on Build GRCh38
rs778241157 Chr.16: 1519949-1519949 on Build GRCh38
rs983693653 Chr.16: 1520012-1520012 on Build GRCh38
rs902428763 Chr.16: 1519926-1519926 on Build GRCh38
rs776809726 Chr.16: 1519963-1519963 on Build GRCh38
rs143385994 Chr.16: 1520009-1520009 on Build GRCh38
rs35823417 Chr.16: 1519928-1519928 on Build GRCh38
rs752465809 Chr.16: 1519849-1519849 on Build GRCh38
rs763121545 Chr.16: 1519994-1519994 on Build GRCh38
rs756795844 Chr.16: 1519868-1519868 on Build GRCh38

View Full Product Details