Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2812656-2812927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITFG2
Entrez Gene ID
55846
Gene Name
integrin alpha FG-GAP repeat containing 2
Gene Aliases
MDS028
UniGene
Hs.446098
Gene Chromosome Location
Chr.12: 2812621-2832217 on Build GRCh38
Transcripts
NM_018463.3 NR_130744.1 XM_005253707.4 XM_005253709.4 XM_017019641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369866 c.89A>G p.N30S Chr.12: 2812849-2812849 on Build GRCh38
6687691 c.79G>T p.D27Y Chr.12: 2812839-2812839 on Build GRCh38
7078578 c.25C>T p.R9C Chr.12: 2812785-2812785 on Build GRCh38
4041286 c.96+2T>C p.? Chr.12: 2812858-2812858 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037764258 Chr.12: 2812892-2812892 on Build GRCh38
rs565678006 Chr.12: 2812774-2812774 on Build GRCh38
rs1021805394 Chr.12: 2812890-2812890 on Build GRCh38
rs987473725 Chr.12: 2812845-2812845 on Build GRCh38
rs940663069 Chr.12: 2812886-2812886 on Build GRCh38
rs747688073 Chr.12: 2812877-2812878 on Build GRCh38
rs764678751 Chr.12: 2812797-2812797 on Build GRCh38
rs761398198 Chr.12: 2812765-2812765 on Build GRCh38
rs960210498 Chr.12: 2812692-2812692 on Build GRCh38
rs753129997 Chr.12: 2812788-2812788 on Build GRCh38
rs777711852 Chr.12: 2812713-2812713 on Build GRCh38
rs764471940 Chr.12: 2812896-2812896 on Build GRCh38
rs746632259 Chr.12: 2812677-2812677 on Build GRCh38
rs369224340 Chr.12: 2812779-2812779 on Build GRCh38
rs7315685 Chr.12: 2812839-2812839 on Build GRCh38
rs745871019 Chr.12: 2812901-2812901 on Build GRCh38
rs188265039 Chr.12: 2812858-2812858 on Build GRCh38
rs113815065 Chr.12: 2812879-2812879 on Build GRCh38
rs750646933 Chr.12: 2812841-2812841 on Build GRCh38
rs375679743 Chr.12: 2812740-2812740 on Build GRCh38
rs536823859 Chr.12: 2812860-2812860 on Build GRCh38
rs771957721 Chr.12: 2812736-2812736 on Build GRCh38
rs527760138 Chr.12: 2812852-2812852 on Build GRCh38
rs770616683 Chr.12: 2812680-2812680 on Build GRCh38
rs775936465 Chr.12: 2812763-2812763 on Build GRCh38
rs1013949128 Chr.12: 2812699-2812699 on Build GRCh38
rs770576465 Chr.12: 2812884-2812884 on Build GRCh38
rs185707858 Chr.12: 2812738-2812738 on Build GRCh38
rs761126495 Chr.12: 2812889-2812889 on Build GRCh38
rs747570304 Chr.12: 2812861-2812861 on Build GRCh38
rs374263180 Chr.12: 2812900-2812900 on Build GRCh38
rs1057080461 Chr.12: 2812848-2812865 on Build GRCh38
rs769282731 Chr.12: 2812766-2812766 on Build GRCh38
rs368219788 Chr.12: 2812737-2812737 on Build GRCh38
rs374716504 Chr.12: 2812866-2812866 on Build GRCh38
rs775776371 Chr.12: 2812885-2812885 on Build GRCh38
rs766719405 Chr.12: 2812907-2812907 on Build GRCh38
rs769325454 Chr.12: 2812863-2812863 on Build GRCh38
rs947536842 Chr.12: 2812709-2812709 on Build GRCh38
rs777169520 Chr.12: 2812899-2812899 on Build GRCh38
rs113509550 Chr.12: 2812826-2812826 on Build GRCh38
rs779988252 Chr.12: 2812856-2812856 on Build GRCh38
rs772882740 Chr.12: 2812770-2812770 on Build GRCh38
rs757366015 Chr.12: 2812717-2812717 on Build GRCh38
rs36050311 Chr.12: 2812878-2812879 on Build GRCh38
rs750595473 Chr.12: 2812904-2812904 on Build GRCh38
rs907885442 Chr.12: 2812874-2812874 on Build GRCh38
rs761022578 Chr.12: 2812790-2812790 on Build GRCh38
rs958242276 Chr.12: 2812746-2812746 on Build GRCh38
rs373369614 Chr.12: 2812867-2812867 on Build GRCh38
rs201298538 Chr.12: 2812802-2812802 on Build GRCh38
rs150848024 Chr.12: 2812827-2812827 on Build GRCh38
rs753959209 Chr.12: 2812715-2812715 on Build GRCh38
rs748936879 Chr.12: 2812876-2812876 on Build GRCh38
rs779746929 Chr.12: 2812906-2812906 on Build GRCh38
rs762644040 Chr.12: 2812777-2812777 on Build GRCh38
rs779222830 Chr.12: 2812727-2812727 on Build GRCh38
rs956282738 Chr.12: 2812684-2812684 on Build GRCh38
rs765390762 Chr.12: 2812903-2812903 on Build GRCh38
rs967157174 Chr.12: 2812718-2812718 on Build GRCh38
rs779169520 Chr.12: 2812836-2812836 on Build GRCh38
rs768418463 Chr.12: 2812859-2812859 on Build GRCh38
rs532036343 Chr.12: 2812729-2812729 on Build GRCh38
rs776271123 Chr.12: 2812705-2812705 on Build GRCh38
rs922239753 Chr.12: 2812785-2812785 on Build GRCh38

View Full Product Details