Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6418800-6419049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES2
Entrez Gene ID
54626
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
bHLHb40
UniGene
Hs.118727
Gene Chromosome Location
Chr.1: 6415232-6419919 on Build GRCh38
Transcripts
NM_019089.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144144 c.415C>T p.P139S Chr.1: 6418980-6418980 on Build GRCh38
6419628 c.400G>A p.G134R Chr.1: 6418995-6418995 on Build GRCh38
6410531 c.497G>A p.G166D Chr.1: 6418898-6418898 on Build GRCh38
5034023 c.353A>C p.H118P Chr.1: 6419042-6419042 on Build GRCh38
6358684 c.515C>T p.P172L Chr.1: 6418880-6418880 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900885746 Chr.1: 6418961-6418961 on Build GRCh38
rs955928303 Chr.1: 6419027-6419027 on Build GRCh38
rs775519801 Chr.1: 6418866-6418866 on Build GRCh38
rs952380380 Chr.1: 6418998-6418998 on Build GRCh38
rs749954154 Chr.1: 6418974-6418974 on Build GRCh38
rs941504068 Chr.1: 6418825-6418825 on Build GRCh38
rs762392716 Chr.1: 6418963-6418963 on Build GRCh38
rs775043603 Chr.1: 6418928-6418928 on Build GRCh38
rs1016560518 Chr.1: 6419004-6419004 on Build GRCh38
rs1012354525 Chr.1: 6418938-6418938 on Build GRCh38
rs1042980004 Chr.1: 6418918-6418918 on Build GRCh38
rs2235687 Chr.1: 6418980-6418980 on Build GRCh38
rs770062454 Chr.1: 6418862-6418862 on Build GRCh38
rs766999135 Chr.1: 6418968-6418968 on Build GRCh38
rs770671660 Chr.1: 6418937-6418954 on Build GRCh38
rs962203107 Chr.1: 6419012-6419012 on Build GRCh38
rs1045552546 Chr.1: 6418966-6418966 on Build GRCh38
rs376955774 Chr.1: 6419003-6419003 on Build GRCh38
rs905738233 Chr.1: 6418979-6418979 on Build GRCh38
rs903014448 Chr.1: 6418925-6418925 on Build GRCh38
rs764519892 Chr.1: 6418916-6418916 on Build GRCh38
rs763436382 Chr.1: 6418887-6418887 on Build GRCh38
rs1019867929 Chr.1: 6418950-6418950 on Build GRCh38
rs766076429 Chr.1: 6418992-6418992 on Build GRCh38
rs12738021 Chr.1: 6419030-6419030 on Build GRCh38
rs918806175 Chr.1: 6418848-6418848 on Build GRCh38
rs896463206 Chr.1: 6418936-6418936 on Build GRCh38
rs1038642564 Chr.1: 6418847-6418847 on Build GRCh38
rs929976888 Chr.1: 6418872-6418872 on Build GRCh38

View Full Product Details