Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57751823-57752094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
UniGene
Hs.275675
Gene Chromosome Location
Chr.16: 57735717-57757250 on Build GRCh38
Transcripts
NM_005886.2 XM_005255772.4 XM_006721121.3 XM_006721122.2 XM_006721123.3 XM_011522810.2 XM_017022860.1 XM_017022861.1 XM_017022862.1 XM_017022863.1 XM_017022864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510553 c.558T>G p.P186P Chr.16: 57751981-57751981 on Build GRCh38
6877311 c.617C>T p.S206F Chr.16: 57752040-57752040 on Build GRCh38
5882738 c.526C>G p.L176V Chr.16: 57751949-57751949 on Build GRCh38
3729651 c.596C>A p.P199H Chr.16: 57752019-57752019 on Build GRCh38
471867 c.615C>T p.A205A Chr.16: 57752038-57752038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782795487 Chr.16: 57752058-57752058 on Build GRCh38
rs959473747 Chr.16: 57751853-57751853 on Build GRCh38
rs782615365 Chr.16: 57751972-57751972 on Build GRCh38
rs370822732 Chr.16: 57752023-57752023 on Build GRCh38
rs782078975 Chr.16: 57752021-57752021 on Build GRCh38
rs782658225 Chr.16: 57751995-57751995 on Build GRCh38
rs782055855 Chr.16: 57752071-57752071 on Build GRCh38
rs781819158 Chr.16: 57751907-57751907 on Build GRCh38
rs782539154 Chr.16: 57751963-57751963 on Build GRCh38
rs373079701 Chr.16: 57752075-57752075 on Build GRCh38
rs782125903 Chr.16: 57752042-57752042 on Build GRCh38
rs782093831 Chr.16: 57751908-57751908 on Build GRCh38
rs1016365576 Chr.16: 57751842-57751842 on Build GRCh38
rs201842513 Chr.16: 57752002-57752002 on Build GRCh38
rs934362365 Chr.16: 57752019-57752019 on Build GRCh38
rs375453942 Chr.16: 57751892-57751892 on Build GRCh38
rs553603033 Chr.16: 57751845-57751845 on Build GRCh38
rs181359252 Chr.16: 57751929-57751929 on Build GRCh38
rs781932266 Chr.16: 57752008-57752008 on Build GRCh38
rs782314799 Chr.16: 57752070-57752070 on Build GRCh38
rs373110632 Chr.16: 57751904-57751904 on Build GRCh38
rs575012509 Chr.16: 57751881-57751881 on Build GRCh38
rs782790218 Chr.16: 57751897-57751897 on Build GRCh38
rs60158050 Chr.16: 57751964-57751964 on Build GRCh38
rs781796817 Chr.16: 57751967-57751967 on Build GRCh38
rs781986158 Chr.16: 57752038-57752038 on Build GRCh38
rs34438625 Chr.16: 57751901-57751901 on Build GRCh38
rs530672883 Chr.16: 57751917-57751917 on Build GRCh38
rs377490029 Chr.16: 57751935-57751935 on Build GRCh38
rs144612958 Chr.16: 57751957-57751957 on Build GRCh38
rs61729343 Chr.16: 57751989-57751989 on Build GRCh38
rs782450704 Chr.16: 57751971-57751971 on Build GRCh38
rs924149287 Chr.16: 57751978-57751978 on Build GRCh38
rs782124773 Chr.16: 57751893-57751893 on Build GRCh38
rs782716081 Chr.16: 57752073-57752073 on Build GRCh38
rs782304206 Chr.16: 57752005-57752005 on Build GRCh38
rs782233425 Chr.16: 57751990-57751990 on Build GRCh38

View Full Product Details