Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89418055-89418308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
UniGene
Hs.252839
Gene Chromosome Location
Chr.10: 89414568-89421002 on Build GRCh38
Transcripts
NM_012420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016517 c.1097G>A p.R366H Chr.10: 89418296-89418296 on Build GRCh38
1968752 c.933G>A p.K311K Chr.10: 89418132-89418132 on Build GRCh38
6683844 c.1041G>A p.M347I Chr.10: 89418240-89418240 on Build GRCh38
428109 c.1028A>G p.D343G Chr.10: 89418227-89418227 on Build GRCh38
295276 c.940C>T p.L314L Chr.10: 89418139-89418139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747404756 Chr.10: 89418196-89418196 on Build GRCh38
rs772821401 Chr.10: 89418204-89418204 on Build GRCh38
rs776597834 Chr.10: 89418261-89418261 on Build GRCh38
rs754532465 Chr.10: 89418077-89418077 on Build GRCh38
rs777262908 Chr.10: 89418204-89418204 on Build GRCh38
rs304448 Chr.10: 89418243-89418243 on Build GRCh38
rs780952714 Chr.10: 89418087-89418087 on Build GRCh38
rs111662059 Chr.10: 89418271-89418271 on Build GRCh38
rs752370089 Chr.10: 89418116-89418116 on Build GRCh38
rs137961574 Chr.10: 89418200-89418200 on Build GRCh38
rs541824745 Chr.10: 89418206-89418206 on Build GRCh38
rs373762041 Chr.10: 89418171-89418171 on Build GRCh38
rs761492728 Chr.10: 89418166-89418166 on Build GRCh38
rs749609057 Chr.10: 89418090-89418090 on Build GRCh38
rs1041589844 Chr.10: 89418253-89418253 on Build GRCh38
rs779379603 Chr.10: 89418235-89418235 on Build GRCh38
rs747028604 Chr.10: 89418251-89418251 on Build GRCh38
rs747098320 Chr.10: 89418157-89418157 on Build GRCh38
rs780773937 Chr.10: 89418246-89418246 on Build GRCh38
rs765034469 Chr.10: 89418167-89418167 on Build GRCh38
rs772734921 Chr.10: 89418119-89418119 on Build GRCh38
rs776952897 Chr.10: 89418161-89418161 on Build GRCh38
rs767044177 Chr.10: 89418205-89418205 on Build GRCh38
rs780908003 Chr.10: 89418085-89418085 on Build GRCh38
rs867721703 Chr.10: 89418140-89418140 on Build GRCh38
rs914362521 Chr.10: 89418213-89418213 on Build GRCh38
rs369610086 Chr.10: 89418113-89418113 on Build GRCh38
rs771362408 Chr.10: 89418108-89418108 on Build GRCh38
rs139979024 Chr.10: 89418283-89418283 on Build GRCh38
rs768832519 Chr.10: 89418255-89418255 on Build GRCh38
rs755743504 Chr.10: 89418222-89418222 on Build GRCh38
rs781378982 Chr.10: 89418284-89418284 on Build GRCh38
rs775644758 Chr.10: 89418137-89418137 on Build GRCh38
rs142354157 Chr.10: 89418238-89418238 on Build GRCh38
rs150755193 Chr.10: 89418197-89418197 on Build GRCh38
rs748364364 Chr.10: 89418268-89418268 on Build GRCh38

View Full Product Details