Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100633250-100633521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG7
Entrez Gene ID
84873
Gene Name
adhesion G protein-coupled receptor G7
Gene Aliases
GPR128
UniGene
Hs.334511
Gene Chromosome Location
Chr.3: 100609589-100696174 on Build GRCh38
Transcripts
NM_032787.2 XM_011513245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035631 c.420T>C p.N140N Chr.3: 100633350-100633350 on Build GRCh38
1035630 c.393A>G p.L131L Chr.3: 100633323-100633323 on Build GRCh38
3584523 c.358T>C p.L120L Chr.3: 100633288-100633288 on Build GRCh38
6511318 c.373C>A p.L125I Chr.3: 100633303-100633303 on Build GRCh38
1327151 c.344C>G p.P115R Chr.3: 100633274-100633274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756417221 Chr.3: 100633399-100633399 on Build GRCh38
rs749508649 Chr.3: 100633405-100633405 on Build GRCh38
rs752320101 Chr.3: 100633275-100633275 on Build GRCh38
rs184858793 Chr.3: 100633389-100633389 on Build GRCh38
rs774589708 Chr.3: 100633416-100633416 on Build GRCh38
rs116327172 Chr.3: 100633307-100633307 on Build GRCh38
rs764908628 Chr.3: 100633428-100633428 on Build GRCh38
rs757772091 Chr.3: 100633385-100633385 on Build GRCh38
rs1039440626 Chr.3: 100633402-100633402 on Build GRCh38
rs968672004 Chr.3: 100633481-100633481 on Build GRCh38
rs768877574 Chr.3: 100633408-100633408 on Build GRCh38
rs757832499 Chr.3: 100633276-100633276 on Build GRCh38
rs566418219 Chr.3: 100633339-100633339 on Build GRCh38
rs774534669 Chr.3: 100633334-100633334 on Build GRCh38
rs1030874880 Chr.3: 100633437-100633437 on Build GRCh38
rs138834427 Chr.3: 100633337-100633337 on Build GRCh38
rs756584697 Chr.3: 100633409-100633411 on Build GRCh38
rs771058341 Chr.3: 100633426-100633426 on Build GRCh38
rs766964340 Chr.3: 100633464-100633464 on Build GRCh38
rs377440433 Chr.3: 100633395-100633395 on Build GRCh38
rs763541878 Chr.3: 100633382-100633382 on Build GRCh38
rs370231378 Chr.3: 100633425-100633425 on Build GRCh38
rs781555773 Chr.3: 100633306-100633306 on Build GRCh38
rs759246633 Chr.3: 100633346-100633346 on Build GRCh38
rs1015200902 Chr.3: 100633461-100633461 on Build GRCh38
rs956213985 Chr.3: 100633454-100633454 on Build GRCh38
rs372793318 Chr.3: 100633429-100633429 on Build GRCh38
rs544404415 Chr.3: 100633458-100633458 on Build GRCh38
rs759016848 Chr.3: 100633447-100633447 on Build GRCh38
rs369393091 Chr.3: 100633285-100633285 on Build GRCh38
rs759424060 Chr.3: 100633427-100633427 on Build GRCh38
rs768591973 Chr.3: 100633324-100633324 on Build GRCh38
rs780367973 Chr.3: 100633403-100633403 on Build GRCh38
rs918505379 Chr.3: 100633327-100633327 on Build GRCh38
rs751020697 Chr.3: 100633384-100633384 on Build GRCh38
rs775566793 Chr.3: 100633316-100633316 on Build GRCh38
rs766342502 Chr.3: 100633468-100633468 on Build GRCh38
rs755041924 Chr.3: 100633455-100633455 on Build GRCh38
rs139753956 Chr.3: 100633379-100633379 on Build GRCh38
rs143646459 Chr.3: 100633286-100633286 on Build GRCh38
rs533780478 Chr.3: 100633349-100633349 on Build GRCh38
rs373484932 Chr.3: 100633387-100633387 on Build GRCh38
rs770055964 Chr.3: 100633309-100633309 on Build GRCh38
rs757720814 Chr.3: 100633297-100633297 on Build GRCh38
rs752268742 Chr.3: 100633350-100633350 on Build GRCh38

View Full Product Details