Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149482566-149482839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDS
Entrez Gene ID
3423
Gene Name
iduronate 2-sulfatase
Gene Aliases
MPS2, SIDS
UniGene
Hs.460960
Gene Chromosome Location
Chr.X: 149476990-149505354 on Build GRCh38
Transcripts
NM_000202.7 NM_001166550.3

SNP Detail

SNP ID Chromosome Location
rs886044837 Chr.X: 149482808-149482808 on Build GRCh38
rs782041174 Chr.X: 149482750-149482750 on Build GRCh38
rs148723108 Chr.X: 149482783-149482783 on Build GRCh38
rs1064458 Chr.X: 149482765-149482765 on Build GRCh38
rs782166680 Chr.X: 149482795-149482795 on Build GRCh38
rs1009125098 Chr.X: 149482684-149482684 on Build GRCh38
rs192118605 Chr.X: 149482709-149482709 on Build GRCh38
rs1021414825 Chr.X: 149482775-149482775 on Build GRCh38
rs890729896 Chr.X: 149482680-149482680 on Build GRCh38
rs145056272 Chr.X: 149482778-149482778 on Build GRCh38
rs368926410 Chr.X: 149482777-149482777 on Build GRCh38
rs188552517 Chr.X: 149482641-149482641 on Build GRCh38
rs781844928 Chr.X: 149482804-149482804 on Build GRCh38
rs782432215 Chr.X: 149482715-149482715 on Build GRCh38
rs782379005 Chr.X: 149482770-149482770 on Build GRCh38
rs782635393 Chr.X: 149482651-149482651 on Build GRCh38
rs782405572 Chr.X: 149482659-149482659 on Build GRCh38
rs782584433 Chr.X: 149482704-149482704 on Build GRCh38
rs782355588 Chr.X: 149482744-149482744 on Build GRCh38
rs782260567 Chr.X: 149482751-149482751 on Build GRCh38
rs993495179 Chr.X: 149482645-149482645 on Build GRCh38
rs975572043 Chr.X: 149482773-149482773 on Build GRCh38
rs1026257602 Chr.X: 149482677-149482677 on Build GRCh38
rs782717602 Chr.X: 149482782-149482782 on Build GRCh38
rs375940553 Chr.X: 149482739-149482739 on Build GRCh38

View Full Product Details