Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15598671-15598920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0430
Entrez Gene ID
9665
Gene Name
KIAA0430
Gene Aliases
LKAP, MARF1, PPP1R34
UniGene
Hs.173524
Gene Chromosome Location
Chr.16: 15594369-15643166 on Build GRCh38
Transcripts
NM_001184998.1 NM_001184999.1 NM_014647.3 XM_005255704.1 XM_005255706.1 XM_005255707.1 XM_017023900.1 XM_017023901.1 XM_017023902.1 XM_017023903.1 XM_017023904.1 XM_017023905.1 XM_017023906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850226 c.4970C>T p.P1657L Chr.16: 15598868-15598868 on Build GRCh38
3706867 c.4031G>A p.R1344H Chr.16: 15598871-15598871 on Build GRCh38
2126789 c.3991G>A p.V1331I Chr.16: 15598911-15598911 on Build GRCh38
4813697 c.4002A>G p.Q1334Q Chr.16: 15598900-15598900 on Build GRCh38
967351 c.4927G>A p.V1643I Chr.16: 15598911-15598911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755976063 Chr.16: 15598776-15598776 on Build GRCh38
rs762678358 Chr.16: 15598821-15598821 on Build GRCh38
rs747503717 Chr.16: 15598851-15598851 on Build GRCh38
rs769060172 Chr.16: 15598864-15598864 on Build GRCh38
rs770123921 Chr.16: 15598890-15598890 on Build GRCh38
rs138588214 Chr.16: 15598829-15598829 on Build GRCh38
rs374459645 Chr.16: 15598810-15598810 on Build GRCh38
rs897281650 Chr.16: 15598894-15598894 on Build GRCh38
rs1035129157 Chr.16: 15598871-15598871 on Build GRCh38
rs915592493 Chr.16: 15598701-15598701 on Build GRCh38
rs562011794 Chr.16: 15598773-15598773 on Build GRCh38
rs776983879 Chr.16: 15598872-15598872 on Build GRCh38
rs941412350 Chr.16: 15598863-15598863 on Build GRCh38
rs777573537 Chr.16: 15598815-15598815 on Build GRCh38
rs1040926576 Chr.16: 15598796-15598796 on Build GRCh38
rs566108863 Chr.16: 15598819-15598819 on Build GRCh38
rs745361071 Chr.16: 15598822-15598822 on Build GRCh38
rs988332659 Chr.16: 15598884-15598884 on Build GRCh38
rs775017207 Chr.16: 15598850-15598850 on Build GRCh38
rs77616598 Chr.16: 15598818-15598818 on Build GRCh38
rs922491260 Chr.16: 15598797-15598797 on Build GRCh38
rs773614564 Chr.16: 15598895-15598895 on Build GRCh38
rs1023632405 Chr.16: 15598719-15598719 on Build GRCh38
rs934471834 Chr.16: 15598792-15598792 on Build GRCh38
rs539162698 Chr.16: 15598800-15598800 on Build GRCh38
rs772894240 Chr.16: 15598770-15598773 on Build GRCh38
rs568895318 Chr.16: 15598772-15598772 on Build GRCh38
rs200228717 Chr.16: 15598900-15598900 on Build GRCh38
rs12445076 Chr.16: 15598793-15598793 on Build GRCh38
rs758090123 Chr.16: 15598815-15598815 on Build GRCh38
rs968838108 Chr.16: 15598708-15598708 on Build GRCh38
rs749043521 Chr.16: 15598816-15598816 on Build GRCh38
rs762148874 Chr.16: 15598883-15598883 on Build GRCh38

View Full Product Details