Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20122617-20122890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT2B
Entrez Gene ID
8850
Gene Name
lysine acetyltransferase 2B
Gene Aliases
CAF, P/CAF, PCAF
UniGene
Hs.533055
Gene Chromosome Location
Chr.3: 20040032-20154404 on Build GRCh38
Transcripts
NM_003884.4 XM_005265528.4 XM_017007423.1 XM_017007424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6347831 c.1374G>T p.M458I Chr.3: 20122765-20122765 on Build GRCh38
4116589 c.1365C>T p.N455N Chr.3: 20122756-20122756 on Build GRCh38
4768972 c.1301C>T p.S434F Chr.3: 20122692-20122692 on Build GRCh38
2948451 c.1337T>G p.M446R Chr.3: 20122728-20122728 on Build GRCh38
2948452 c.1366G>A p.E456K Chr.3: 20122757-20122757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149881135 Chr.3: 20122756-20122756 on Build GRCh38
rs575172601 Chr.3: 20122766-20122766 on Build GRCh38
rs1003611326 Chr.3: 20122788-20122788 on Build GRCh38
rs755332489 Chr.3: 20122777-20122777 on Build GRCh38
rs776596621 Chr.3: 20122831-20122831 on Build GRCh38
rs749392601 Chr.3: 20122792-20122792 on Build GRCh38
rs368666068 Chr.3: 20122665-20122665 on Build GRCh38
rs144806388 Chr.3: 20122683-20122683 on Build GRCh38
rs763841855 Chr.3: 20122812-20122812 on Build GRCh38
rs979990403 Chr.3: 20122864-20122864 on Build GRCh38
rs747506090 Chr.3: 20122721-20122721 on Build GRCh38
rs928537315 Chr.3: 20122865-20122865 on Build GRCh38
rs756765231 Chr.3: 20122759-20122759 on Build GRCh38
rs763865060 Chr.3: 20122829-20122829 on Build GRCh38
rs753739665 Chr.3: 20122758-20122758 on Build GRCh38
rs780458718 Chr.3: 20122711-20122711 on Build GRCh38
rs9310610 Chr.3: 20122838-20122838 on Build GRCh38
rs771703439 Chr.3: 20122724-20122724 on Build GRCh38
rs969391957 Chr.3: 20122849-20122849 on Build GRCh38
rs778576070 Chr.3: 20122689-20122689 on Build GRCh38
rs774743242 Chr.3: 20122796-20122796 on Build GRCh38
rs779697857 Chr.3: 20122772-20122772 on Build GRCh38
rs763307456 Chr.3: 20122752-20122752 on Build GRCh38
rs761915404 Chr.3: 20122732-20122732 on Build GRCh38
rs772390952 Chr.3: 20122695-20122695 on Build GRCh38
rs767417068 Chr.3: 20122753-20122753 on Build GRCh38
rs144875299 Chr.3: 20122776-20122776 on Build GRCh38
rs777585016 Chr.3: 20122668-20122668 on Build GRCh38
rs553434368 Chr.3: 20122760-20122760 on Build GRCh38
rs373525362 Chr.3: 20122755-20122755 on Build GRCh38
rs201839892 Chr.3: 20122757-20122757 on Build GRCh38
rs759916964 Chr.3: 20122828-20122828 on Build GRCh38
rs758109228 Chr.3: 20122769-20122769 on Build GRCh38
rs767470394 Chr.3: 20122646-20122646 on Build GRCh38
rs377475879 Chr.3: 20122738-20122738 on Build GRCh38
rs748653359 Chr.3: 20122785-20122785 on Build GRCh38
rs756747536 Chr.3: 20122684-20122684 on Build GRCh38
rs577306981 Chr.3: 20122740-20122740 on Build GRCh38
rs760495333 Chr.3: 20122799-20122799 on Build GRCh38
rs781690402 Chr.3: 20122783-20122783 on Build GRCh38
rs886934542 Chr.3: 20122806-20122806 on Build GRCh38
rs41285059 Chr.3: 20122790-20122790 on Build GRCh38
rs199552217 Chr.3: 20122653-20122653 on Build GRCh38
rs1030559923 Chr.3: 20122733-20122733 on Build GRCh38
rs777011461 Chr.3: 20122728-20122728 on Build GRCh38
rs771209664 Chr.3: 20122793-20122793 on Build GRCh38
rs114528005 Chr.3: 20122664-20122664 on Build GRCh38
rs773232310 Chr.3: 20122791-20122791 on Build GRCh38
rs146721823 Chr.3: 20122692-20122692 on Build GRCh38

View Full Product Details